Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04278301_m1
          See other E030013I19RIK GE Assays ›
          Gene Symbol:
          E030013I19Rik
          Gene Name:
          RIKEN cDNA E030013I19 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 12301246 - 12312315 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04278301_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: E030013I19Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040353.1 1 - 2 342 119
          GenBank mRNA AK086420.1 - 1 - 2 249 119
          AK142619.1 - 1 - 2 269 119
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB663013.1 - 1 - 2 249 Chr. 2 - 12301340 - 12310254 [+] on Build GRCm38 119
          GenBankmRNA BY730798.1 - 1 - 2 269 Chr. 2 - 12301319 - 12310265 [+] on Build GRCm38 119

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream integrin alpha 8 Itga8 AI447669 Anti-sense Chr. 2 - 12106632 - 12301996 on NCBI Build GRCm38
          Downstream family with sequence similarity 188,
          member A
          Fam188a 1810041E18Rik,
          2310047O13Rik,
          5830410F13Rik,
          AI447827,
          AW111958
          34.9kb Anti-sense Chr. 2 - 12347263 - 12419490 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000225.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2253607 AK142619.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 386719 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 861925 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 13151 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 214868 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 591004 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 92843 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139846 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2253607 AK142619.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139846 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111334 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21010340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21010353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21010349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01495649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21010344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01812507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21010342 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01521422 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21010343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21010358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01846849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21010359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21010339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21010348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21010357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01705853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01521419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21010356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21010345 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200000909.mm.1 TC0200000225.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200000909.mm.1 NR_040353.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000909.mm.1 NR_040353.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000911.mm.1 TC0200000225.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200000911.mm.1 NR_040353.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000911.mm.1 NR_040353.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000912.mm.1 TC0200000225.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200000912.mm.1 NR_040353.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000912.mm.1 NR_040353.1 Refseq
          Mouse Expression Set 430 1447490_at - -
          Mouse Expression Set 430 1447490_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 538738 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0