Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04278348_m1
          See other 3110045C21RIK GE Assays ›
          Gene Symbol:
          3110045C21Rik
          Gene Name:
          RIKEN cDNA 3110045C21 gene
          Gene Aliases:
          AI838577
          Chromosome Location:
          Chr.1: 169969409 - 169972396 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04278348_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 3110045C21Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040438.1 1 - 2 168 138
          GenBank mRNA AK014177.1 - 1 - 2 169 138
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BM939629.1 - 1 - 2 95 Chr. 1 - 169969482 - 169971971 [+] on Build GRCm38 138
          GenBankmRNA CA494857.1 - 1 - 2 163 Chr. 1 - 169969449 - 169972056 [+] on Build GRCm38 138
          GenBankmRNA BY713148.1 - 1 - 2 169 Chr. 1 - 169969409 - 169972120 [+] on Build GRCm38 138

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream hydroxysteroid (17-beta) dehydrogenase 7 Hsd17b7 AI266814,
          ERG27
          -171.0 Anti-sense Chr. 1 - 169949535 - 169969238 on NCBI Build GRCm38
          Downstream discoidin domain receptor family,
          member 2
          Ddr2 AW495251,
          Ntrkr3,
          tyro10
          1.5kb Anti-sense Chr. 1 - 169972307 - 170110836 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001564.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 841742 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 261983 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 76515 - -
          Whole Mouse Genome G4122A A_52_P296864 AK014177.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P296864 AK014032.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129595 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1430401_PM_at - -
          Mouse Genome 430 2.0 Array 1430401_PM_at - -
          Mouse Genome 430 2.0 Array 1430401_at - -
          Affymetrix Mouse Clariom S Array PSR0100012820.mm.1 TC0100001564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100012820.mm.1 NR_040438.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012820.mm.1 NR_040438.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012822.mm.1 TC0100001564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100012822.mm.1 NR_040438.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012822.mm.1 NR_040438.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012816.mm.1 TC0100001564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100012816.mm.1 NR_040438.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012816.mm.1 NR_040438.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012821.mm.1 TC0100001564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100012821.mm.1 NR_040438.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012821.mm.1 NR_040438.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012819.mm.1 TC0100001564.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100012819.mm.1 NR_040438.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100012819.mm.1 NR_040438.1 Refseq
          Mouse Expression Set 430 1430401_PM_at - -
          Mouse Expression Set 430 1430401_at - -
          Murine Genome U74v2 Set 130675_at - -
          Murine Genome U74v2 Set 130675_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430401_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430401_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 316559 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100670575 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0