Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04278359_m1
          See other GM10548 GE Assays ›
          Gene Symbol:
          Gm10548
          Gene Name:
          ribosomal protein L29 pseudogene
          Gene Aliases:
          ENSMUSG00000073607
          Chromosome Location:
          Chr.18: 34207775 - 34221772 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04278359_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm10548
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040534.1 1 - 2 294 67
          GenBank mRNA AK160850.1 - 1 - 2 296 67
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY713558.1 - 1 - 2 296 Chr. 18 - 34209712 - 34221772 [-] on Build GRCm38 67
          GenBankmRNA BY122674.1 - 1 - 2 174 Chr. 18 - 34209971 - 34221651 [-] on Build GRCm38 67

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream erythrocyte protein band 4.1 like 4a Epb4.1l4a Epb4.1l4,
          Epb41l4a,
          NBL4
          -200.1kb Sense Chr. 18 - 33796327 - 34007678 on NCBI Build GRCm38
          Downstream signal recognition particle 19 Srp19 2310020D23Rik 109.3kb Anti-sense Chr. 18 - 34331121 - 34336599 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001146.mm.1
          This assay targets the same location as RTA transcript cluster(s) TC1800001173.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2091874 AK132217.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2091556 AK132217.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 918441 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 838493 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 952389 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 603080 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 41328 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 455244 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 718228 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 13751 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 163978 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 150999 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P105771 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2091556 AK132217.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2091874 AK132217.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P31611512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31611481 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31611538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31611540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13676807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13772172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31611504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31611476 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31611526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31611513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31611472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14110389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31611547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31611548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31611508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31611529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31611500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13676810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31611502 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31611524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31611536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31611521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31611519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31611543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14009764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31611474 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31611514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31611523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31611528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31611509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31611479 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10458087 - -
          Affymetrix Mouse Gene 1.1 ST Array 10458086 - -
          Affymetrix Mouse Clariom S Array PSR1800008347.mm.1 TC1800001146.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008347.mm.1 NR_040534.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008347.mm.1 NR_040534.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008348.mm.1 TC1800001146.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008348.mm.1 NR_040534.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008348.mm.1 NR_040534.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008349.mm.1 TC1800001146.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008349.mm.1 NR_040534.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008349.mm.1 NR_040534.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008350.mm.1 TC1800001146.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008350.mm.1 NR_040534.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008350.mm.1 NR_040534.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0