Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04278367_m1
          See other 2010009K17RIK GE Assays ›
          Gene Symbol:
          2010009K17Rik
          Gene Name:
          RIKEN cDNA 2010009K17 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 158070364 - 158091797 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04278367_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2010009K17Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040609.1 3 - 4 294 81
          GenBank mRNA AK008166.1 - 3 - 4 298 81
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY708124.1 - 3 - 4 297 Chr. 2 - 158070364 - 158091797 [-] on Build GRCm38 81

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream TELO2 interacting protein 1 Tti1 2610036D13Rik,
          AI449463
          -41.9kb Sense Chr. 2 - 157981803 - 158028477 on NCBI Build GRCm38
          Downstream transglutaminase 2,
          C polypeptide
          Tgm2 G[a]h,
          TG2,
          TGase2,
          tTG,
          tTGas
          32.3kb Sense Chr. 2 - 158116405 - 158146392 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200005029.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02207771 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02114241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02074286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21780934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02234758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01936685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21780905 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21780943 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21780912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21780927 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21780940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21780932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01873033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01936683 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02114233 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21780908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21780906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21780911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21780939 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01763841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02114244 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21780941 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21780918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21780946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02114238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02114231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01299641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21780904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01873032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21780947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21780938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02114228 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21780957 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01873031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21780922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21780903 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02114246 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01580871 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21780913 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200043041.mm.1 TC0200005029.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200043041.mm.1 NR_040609.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043041.mm.1 NR_040609.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 719670 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106900100 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0