Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04278565_m1
          See other 1700012D01RIK GE Assays ›
          Gene Symbol:
          1700012D01Rik
          Gene Name:
          RIKEN cDNA 1700012D01 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 127667123 - 127668851 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04278565_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700012D01Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045171.1 1 - 2 264 84
          GenBank mRNA CA494581.1 - 1 - 2 267 84
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BU937250.1 - 263 Chr. - - [-] on Build 84
          GenBankmRNA CA466680.1 - 190 Chr. - - [-] on Build 84
          GenBankmRNA BY709030.1 - 1 - 2 258 Chr. 10 - 127667135 - 127668851 [+] on Build GRCm38 84

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene 16230 Gm16230 OTTMUSG00000031895 Anti-sense Chr. 10 - 127668493 - 127676759 on NCBI Build GRCm38
          Downstream transmembrane protein 194 Tmem194 Tmem194a 8.2kb Sense Chr. 10 - 127677065 - 127701049 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000001554.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1459813_PM_at - -
          Mouse Genome 430 2.0 Array 1459813_PM_at - -
          Mouse Genome 430 2.0 Array 1459813_at - -
          Affymetrix Mouse Clariom S Array PSR1000011038.mm.1 TC1000001554.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000011038.mm.1 NR_045171.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011038.mm.1 NR_045171.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011040.mm.1 TC1000001554.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000011040.mm.1 NR_045171.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011040.mm.1 NR_045171.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011041.mm.1 TC1000001554.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000011041.mm.1 NR_045171.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011041.mm.1 NR_045171.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011035.mm.1 TC1000001554.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000011035.mm.1 NR_045171.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000011035.mm.1 NR_045171.1 Refseq
          Mouse Expression Set 430 1459813_PM_at - -
          Mouse Expression Set 430 1459813_at - -
          Murine Genome U74v2 Set 167507_at - -
          Murine Genome U74v2 Set 167507_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1459813_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1459813_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 664213 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0