Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04278648_m1
          See other GM10433 GE Assays ›
          Gene Symbol:
          Gm10433
          Gene Name:
          predicted gene 10433
          Gene Aliases:
          ENSMUSG00000072884
          Chromosome Location:
          Chr.12: 100187968 - 100193538 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04278648_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm10433
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045282.1 2 - 3 479 96
          GenBank mRNA AK141725.1 - 2 - 3 480 96
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CJ064275.1 - 2 - 3 480 Chr. 12 - 100187968 - 100192205 [+] on Build GRCm38 96
          GenBankmRNA DV662061.1 - 2 - 3 131 Chr. 12 - 100188773 - 100192741 [+] on Build GRCm38 96

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream nrde-2 necessary for RNA interference,
          domain containing
          Nrde2 6720454P05Rik -29.0kb Anti-sense Chr. 12 - 100125450 - 100159931 on NCBI Build GRCm38
          Downstream calmodulin 1 Calm1 AI256814,
          AI327027,
          AI461935,
          AL024000,
          CaM,
          Calm
          6.0kb Sense Chr. 12 - 100199541 - 100209806 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200000993.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK141725.1 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2062558 AK141725.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2062558 AK141725.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02588921 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02588930 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02588930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09935876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680445 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09935874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680428 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680450 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680447 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680467 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680439 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09997726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09973655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680441 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680452 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680444 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680473 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09908176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680495 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680458 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09856375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680465 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09849037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680492 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680453 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680446 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09908179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680449 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680454 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680429 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680464 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09997725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680443 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28680461 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200006926.mm.1 TC1200000993.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200006926.mm.1 NR_045282.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200006926.mm.1 NR_045282.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200006927.mm.1 TC1200000993.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200006927.mm.1 NR_045282.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200006927.mm.1 NR_045282.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline