Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04278698_m1
          See other GM13582 GE Assays ›
          Gene Symbol:
          Gm13582
          Gene Name:
          predicted gene 13582
          Gene Aliases:
          ENSMUSG00000075325, OTTMUSG00000012844
          Chromosome Location:
          Chr.2: 60964376 - 60975151 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04278698_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm13582
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045335.1 3 - 4 1197 92
          GenBank mRNA AK138134.1 - 3 - 4 1198 92

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RNA binding motif,
          single stranded interacting protein 1
          Rbms1 2600014B10Rik,
          AI255215,
          MSSP-1,
          MSSP-2,
          MSSP-3,
          YC1
          -926.0 Sense Chr. 2 - 60750193 - 60963450 on NCBI Build GRCm38
          Downstream TRAF family member-associated Nf-kappa B activator Tank C86182,
          E430026L09Rik,
          I-TRAF
          610.4kb Anti-sense Chr. 2 - 61578561 - 61654171 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003617.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2060474 AK138134.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111048 AK138134.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 214449 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 424221 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 589472 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 691138 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701819 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 43712 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175766 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111048 AK138134.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2060474 AK138134.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175766 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01241985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261827 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261834 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261798 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261809 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261833 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261861 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261804 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261794 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01404496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01241982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01248382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02083415 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02015269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261835 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261808 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21261831 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200030866.mm.1 TC0200003617.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200030866.mm.1 NR_045335.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030866.mm.1 NR_045335.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-ffb8c786-92mpb:80/100.66.129.117:80.
          git-commit: 1a801765356e65be0b6ed8595ed79b52bb698fed
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0