Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04335100_mH
          See other A230056P14RIK GE Assays ›
          Gene Symbol:
          A230056P14Rik
          Gene Name:
          RIKEN cDNA A230056P14 gene
          Gene Aliases:
          OTTMUSG00000027345
          Chromosome Location:
          Chr.7: 55962531 - 55980096 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04335100_mH
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A230056P14Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015495.2 5 - 6 746 93
          GenBank mRNA AK043630.1 - 5 - 6 747 93
          AK075669.1 - 89 93
          AK140288.1 - 5 - 6 747 93
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB609724.1 - 1 - 2 89 Chr. 7 - 55976903 - 55980004 [+] on Build GRCm38 93

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream non imprinted in Prader-Willi/Angelman syndrome 1 homolog (human) Nipa1 1110027G09Rik,
          A830014A18Rik,
          FSP3,
          Spg6
          Anti-sense Chr. 7 - 55978484 - 56019573 on NCBI Build GRCm38
          Downstream hect (homologous to the E6-AP (UBE3A) carboxyl terminus) domain and RCC1 (CHC1)-like domain (RLD) 2 Herc2 D15F32S1h,
          D7H15F32S1,
          D7H15F37S1,
          jdf2,
          mKIAA0393,
          rjs
          70.1kb Sense Chr. 7 - 56050155 - 56231800 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000934.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK140288.1 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 459509 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 508390 AB212223.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 508390 AK081203.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 508390 NM_001037907.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 818998 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 569887 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584896 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 851356 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 151877 - -
          Whole Mouse Genome G4122A A_51_P279666 AK140288.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P279666 AK043630.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P279666 AK075669.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P279666 NR_015495.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139406 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10553638 - -
          Affymetrix Mouse Gene 1.0 ST Array 10553636 - -
          Affymetrix Mouse Gene 1.0 ST Array 10553640 - -
          Affymetrix Mouse Gene 1.0 ST Array 10553637 - -
          Affymetrix Mouse Gene 1.0 ST Array 10553639 - -
          Affymetrix HT MG-430 PM Array Plate 1438965_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10563852 - -
          Affymetrix Mouse Gene 1.1 ST Array 10553635 - -
          Mouse Genome 430 2.0 Array 1438965_PM_x_at - -
          Mouse Genome 430 2.0 Array 1438965_x_at - -
          Affymetrix Mouse Clariom S Array PSR0700008503.mm.1 TC0700000934.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700008503.mm.1 NR_015495.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700008501.mm.1 TC0700000934.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700008501.mm.1 NR_015495.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700008502.mm.1 TC0700000934.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700008502.mm.1 NR_015495.2 Refseq
          Mouse Expression Set 430 1438965_PM_x_at - -
          Mouse Expression Set 430 1438965_x_at - -
          Murine Genome U74v2 Set 138508_PM_at - -
          Murine Genome U74v2 Set 138508_at - -
          Affymetrix HT Mouse Genome 430B Array 1438965_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1438965_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 734641 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0