Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04335112_m1
          See other A730017L22RIK GE Assays ›
          Gene Symbol:
          A730017L22Rik
          Gene Name:
          RIKEN cDNA A730017L22 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 130872541 - 130906396 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04335112_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A730017L22Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015523.2 1 - 2 185 96
          GenBank mRNA AK158456.1 - 1 - 2 186 96
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CJ142577.1 - 1 - 2 186 Chr. 2 - 130874293 - 130906396 [-] on Build GRCm38 96

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 4930402H24 gene 4930402H24Rik 2810487F15Rik,
          AI413763,
          C78926
          -34.7kb Sense Chr. 2 - 130706200 - 130840807 on NCBI Build GRCm38
          Downstream attractin Atrn AW558010,
          Mgca,
          mKIAA0548,
          mahogany,
          mg
          51.0 Anti-sense Chr. 2 - 130906447 - 131030333 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200005506.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200039779.mm.1 TC0200005506.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039779.mm.1 NR_015523.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039781.mm.1 TC0200005506.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039781.mm.1 NR_015523.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039788.mm.1 TC0200005506.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039788.mm.1 NR_015523.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039805.mm.1 TC0200005506.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039805.mm.1 NR_015523.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039808.mm.1 TC0200005506.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039808.mm.1 NR_015523.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039807.mm.1 TC0200005506.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039807.mm.1 NR_015523.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039810.mm.1 TC0200005506.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039810.mm.1 NR_015523.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039780.mm.1 TC0200005506.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039780.mm.1 NR_015523.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039786.mm.1 TC0200005506.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039786.mm.1 NR_015523.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039818.mm.1 TC0200005506.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039818.mm.1 NR_015523.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039782.mm.1 TC0200005506.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039782.mm.1 NR_015523.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039783.mm.1 TC0200005506.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039783.mm.1 NR_015523.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0