Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04335389_m1
          See other D5ERTD605E GE Assays ›
          Gene Symbol:
          D5Ertd605e
          Gene Name:
          DNA segment, Chr 5, ERATO Doi 605, expressed
          Gene Aliases:
          Gm2433
          Chromosome Location:
          Chr.5: 147418620 - 147423044 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04335389_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: D5Ertd605e
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033625.1 2 - 3 253 102
          GenBank mRNA AK038337.1 - 2 - 3 254 102
          BG069957.2 - 1 - 2 102
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB631808.1 - 2 - 3 254 Chr. 5 - 147418620 - 147422610 [+] on Build GRCm38 102

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream FMS-like tyrosine kinase 3 Flt3 B230315G04,
          CD135,
          Flk-2,
          Flk2,
          Flt-3,
          Ly72,
          wmfl
          -20.1kb Anti-sense Chr. 5 - 147330741 - 147400489 on NCBI Build GRCm38
          Downstream PAN3 polyA specific ribonuclease subunit homolog (S. cerevisiae) Pan3 2700050F09Rik,
          A430027N15Rik,
          AU014670
          7.1kb Sense Chr. 5 - 147430162 - 147548502 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001807.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 134546 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 2887 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 12707 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 2743 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174106 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174106 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04858367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04164874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04659341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04605519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04914416 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04845586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24173564 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04659342 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500016690.mm.1 TC0500001807.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500016690.mm.1 NR_033625.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016690.mm.1 NR_033625.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016687.mm.1 TC0500001807.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500016687.mm.1 NR_033625.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016687.mm.1 NR_033625.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016689.mm.1 TC0500001807.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500016689.mm.1 NR_033625.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016689.mm.1 NR_033625.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016684.mm.1 TC0500001807.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500016684.mm.1 NR_033625.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016684.mm.1 NR_033625.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016691.mm.1 TC0500001807.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500016691.mm.1 NR_033625.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500016691.mm.1 NR_033625.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0