Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04335453_m1
          See other 1810018F18RIK GE Assays ›
          Gene Symbol:
          1810018F18Rik
          Gene Name:
          RIKEN cDNA 1810018F18 gene
          Gene Aliases:
          1810031L05Rik, 1810047O18Rik
          Chromosome Location:
          Chr.19: 58698917 - 58701323 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04335453_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1810018F18Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038140.1 2 - 3 70 156
          GenBank mRNA AK007531.1 - 2 - 3 70 156
          AK007668.1 - 2 - 3 73 156

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 1810035K13 gene 1810035K13Rik -2.9kb Sense Chr. 19 - 58695506 - 58696178 on NCBI Build GRCm38
          Downstream pancreatic lipase related protein 1 Pnliprp1 Plrp1 27.6kb Sense Chr. 19 - 58728887 - 58744169 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000840.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 687686 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 68888 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14150974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32214787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32214792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32214795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32214794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14150969 - -
          Array Name Probe IDs Targets Target Type
          Mouse Genome 430 2.0 Array 1453870_PM_at - -
          Mouse Genome 430 2.0 Array 1453870_at - -
          Affymetrix Mouse Clariom S Array PSR1900007294.mm.1 TC1900000840.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900007294.mm.1 NR_038140.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007294.mm.1 NR_038140.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007296.mm.1 TC1900000840.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900007296.mm.1 NR_038140.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007296.mm.1 NR_038140.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007293.mm.1 TC1900000840.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900007293.mm.1 NR_038140.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007293.mm.1 NR_038140.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 710138 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0