Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04335481_m1
          See other 1700025B11RIK GE Assays ›
          Gene Symbol:
          1700025B11Rik
          Gene Name:
          RIKEN cDNA 1700025B11 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 77558239 - 77561892 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04335481_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700025B11Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040310.1 1 - 2 68 113
          GenBank mRNA AK006321.1 - 1 - 2 39 113
          CN841536.1 - 71 113
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY706394.1 - 1 - 2 38 Chr. 15 - 77558242 - 77561862 [-] on Build GRCm38 113

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream apolipoprotein L 7c Apol7c 2210421G13Rik -25.8kb Sense Chr. 15 - 77524852 - 77533335 on NCBI Build GRCm38
          Downstream apolipoprotein L 10B Apol10b 9130218O11Rik 22.2kb Sense Chr. 15 - 77584106 - 77598542 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001813.mm.1
          This assay targets the same location as RTA transcript cluster(s) TC0700001397.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_03_of_03_v2 A_68_P30354657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12017853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30354645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30354654 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30354652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11896671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30354655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11956009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30354622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11944472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30354640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30354663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30354659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30354642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30354662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12400100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30354666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30354660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30354668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30354651 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1500014078.mm.1 TC1500001813.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500014078.mm.1 NR_040310.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014078.mm.1 NR_040310.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014075.mm.1 TC1500001813.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500014075.mm.1 NR_040310.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014075.mm.1 NR_040310.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014076.mm.1 TC1500001813.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500014076.mm.1 NR_040310.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014076.mm.1 NR_040310.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014077.mm.1 TC1500001813.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500014077.mm.1 NR_040310.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014077.mm.1 NR_040310.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 532486 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6980136 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b7b59cd5b-z8v7b:80/100.66.130.62:80.
          git-commit: 87deb889a5bb48f859ebd9970e87be562fd9396c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0