Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04335704_m1
          See other CSF3R GE Assays ›
          Gene Symbol:
          Csf3r
          Gene Name:
          colony stimulating factor 3 receptor (granulocyte)
          Gene Aliases:
          Cd114, Csfgr, G-CSF-R, G-CSFR
          Chromosome Location:
          Chr.4: 126023441 - 126044975 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04335704_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Csf3r
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045561.1 9 - 10 1458 97
          GenBank mRNA AK144318.1 - 9 - 10 1458 97

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 2610028E06 gene 2610028E06Rik BB043107 -113.5kb Anti-sense Chr. 4 - 125887851 - 125922310 on NCBI Build GRCm38
          Downstream mitochondrial ribosomal protein S15 Mrps15 1500003E24Rik,
          2410002B11Rik,
          Mprs15
          9.2kb Sense Chr. 4 - 126046928 - 126055536 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001449.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          neutrophil chemotaxis
          regulation of myeloid cell differentiation
          cytokine receptor activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400011006.mm.1 TC0400001449.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011006.mm.1 NM_001252651.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011006.mm.1 NM_001252651.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011006.mm.1 NM_007782.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011006.mm.1 NM_007782.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011006.mm.1 NR_045561.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011006.mm.1 NR_045561.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011007.mm.1 TC0400001449.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011007.mm.1 NM_001252651.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011007.mm.1 NM_001252651.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011007.mm.1 NM_007782.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011007.mm.1 NM_007782.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011007.mm.1 NR_045561.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011007.mm.1 NR_045561.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011013.mm.1 TC0400001449.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011013.mm.1 NM_001252651.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011013.mm.1 NM_001252651.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011013.mm.1 NM_007782.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011013.mm.1 NM_007782.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011013.mm.1 NR_045561.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011013.mm.1 NR_045561.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011016.mm.1 TC0400001449.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011016.mm.1 NM_001252651.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011016.mm.1 NM_001252651.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011016.mm.1 NM_007782.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011016.mm.1 NM_007782.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011016.mm.1 NR_045561.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011016.mm.1 NR_045561.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011009.mm.1 TC0400001449.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011009.mm.1 NM_001252651.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011009.mm.1 NM_001252651.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011009.mm.1 NM_007782.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011009.mm.1 NM_007782.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011009.mm.1 NR_045561.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011009.mm.1 NR_045561.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011008.mm.1 TC0400001449.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011008.mm.1 NM_001252651.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011008.mm.1 NM_001252651.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011008.mm.1 NM_007782.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011008.mm.1 NM_007782.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011008.mm.1 NR_045561.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011008.mm.1 NR_045561.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011011.mm.1 TC0400001449.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011011.mm.1 NM_001252651.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011011.mm.1 NM_001252651.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011011.mm.1 NM_007782.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011011.mm.1 NM_007782.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011011.mm.1 NR_045561.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011011.mm.1 NR_045561.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011004.mm.1 TC0400001449.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011004.mm.1 NR_045561.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011004.mm.1 NR_045561.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011012.mm.1 TC0400001449.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011012.mm.1 NM_001252651.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011012.mm.1 NM_001252651.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011012.mm.1 NM_007782.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011012.mm.1 NM_007782.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011012.mm.1 NR_045561.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011012.mm.1 NR_045561.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011014.mm.1 TC0400001449.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011014.mm.1 NM_001252651.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011014.mm.1 NM_001252651.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011014.mm.1 NM_007782.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011014.mm.1 NM_007782.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011014.mm.1 NR_045561.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011014.mm.1 NR_045561.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011005.mm.1 TC0400001449.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011005.mm.1 NM_001252651.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011005.mm.1 NM_001252651.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011005.mm.1 NM_007782.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011005.mm.1 NM_007782.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011005.mm.1 NR_045561.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011005.mm.1 NR_045561.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011010.mm.1 TC0400001449.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011010.mm.1 NM_001252651.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011010.mm.1 NM_001252651.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011010.mm.1 NM_007782.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011010.mm.1 NM_007782.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011010.mm.1 NR_045561.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011010.mm.1 NR_045561.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 735450 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0