Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04335925_m1
          See other 1700066N21RIK GE Assays ›
          Gene Symbol:
          1700066N21Rik
          Gene Name:
          RIKEN cDNA 1700066N21 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 87908588 - 87979351 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04335925_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700066N21Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045924.1 4 - 5 574 69
          GenBank mRNA AK006908.1 - 4 - 5 575 69
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY706904.1 - 4 - 5 579 Chr. 5 - 87908583 - 87979351 [-] on Build GRCm38 69

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream cDNA sequence BC037156 BC037156 FDC-SP Anti-sense Chr. 5 - 87908779 - 87914171 on NCBI Build GRCm38
          Downstream casein kappa Csn3 AW208918,
          CSN10,
          Csnk
          13.2kb Anti-sense Chr. 5 - 87925575 - 87932545 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002705.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 353555 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 25042 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 296994 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 288063 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 915729 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 122765 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 760505 AK016084.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 476628 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 888532 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 628804 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 896141 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 935793 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 876105 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 727966 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 591482 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 241508 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 251289 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 65404 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 833040 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 960048 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 733083 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 377977 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 367089 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 437625 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 215580 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 489995 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 254144 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 667588 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 6648 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 82012 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 178300 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P135807 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115895 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145288 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05645939 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05645872 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05646027 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01154596 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05645894 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P135807 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05645958 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05645944 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01154641 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P145288 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115895 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05645886 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P135807 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P145288 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115895 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05645958 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05645872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870209 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870204 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870210 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870193 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870217 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870192 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870197 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04969823 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870199 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870206 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500023883.mm.1 TC0500002705.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500023883.mm.1 NR_045924.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023883.mm.1 NR_045924.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023884.mm.1 TC0500002705.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500023884.mm.1 NR_045924.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023884.mm.1 NR_045924.1 Refseq
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105050184 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6bbccd986b-mx52t:80/100.66.130.62:80.
          git-commit: 717389b07c2b9f75290ff02e811d41bb86d4254a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0