Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04335951_m1
          See other GM10373 GE Assays ›
          Gene Symbol:
          Gm10373
          Gene Name:
          predicted gene 10373
          Gene Aliases:
          ENSMUSG00000072592
          Chromosome Location:
          Chr.15: 43430943 - 43477036 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04335951_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm10373
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046064.1 2 - 3 259 78
          GenBank mRNA AK148391.1 - 2 - 3 260 78
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY479926.1 - 1 - 2 127 Chr. 15 - 43430943 - 43439697 [-] on Build GRCm38 78
          GenBankmRNA BY741520.1 - 2 - 3 260 Chr. 15 - 43430945 - 43477036 [-] on Build GRCm38 78

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream eukaryotic translation initiation factor 3,
          subunit E
          Eif3e 48kDa,
          Eif3s6,
          Int6,
          eIF3-p46,
          eIF3-p48
          -148.2kb Sense Chr. 15 - 43250040 - 43282736 on NCBI Build GRCm38
          Downstream ER membrane protein complex subunit 2 Emc2 4921531G14Rik,
          AV060620,
          AW209495,
          Ttc35
          37.4kb Anti-sense Chr. 15 - 43477214 - 43527777 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001461.mm.1
          This assay targets the same location as RTA transcript cluster(s) TC0700003072.rn.1

          Additional Information:

          This assay is in the 5'UTR region of the AK148391.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P132491 AK148391.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115645 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P132491 AK148391.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08351302 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08351336 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115645 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08351322 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08351302 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08351336 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176237 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176231 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12039947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12069212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11882024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176223 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12432108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12069191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12131691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176109 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12069194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176222 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176093 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30176197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11981881 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1500011153.mm.1 TC1500001461.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500011153.mm.1 NR_046064.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500011153.mm.1 NR_046064.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500011155.mm.1 TC1500001461.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500011155.mm.1 NR_046064.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500011155.mm.1 NR_046064.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500011154.mm.1 TC1500001461.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500011154.mm.1 NR_046064.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500011154.mm.1 NR_046064.1 Refseq
          Murine Genome U74v2 Set 133961_PM_at - -
          Murine Genome U74v2 Set 133961_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0