Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04336131_sH
          See other 1700084J12RIK GE Assays ›
          Gene Symbol:
          1700084J12Rik
          Gene Name:
          ribosomal protein L7-like 1 pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 33405209 - 33405939 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04336131_sH
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700084J12Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033608.1 1 677 107
          GenBank mRNA AK006993.1 - 1 715 107

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream carboxypeptidase Q Cpq 1190003P12Rik,
          2610034C17Rik,
          Hls2,
          Lal-1,
          Pgcp
          Anti-sense Chr. 15 - 33083129 - 33594552 on NCBI Build GRCm38
          Downstream RIKEN cDNA 4930592A05 gene 4930592A05Rik 187.9kb Sense Chr. 15 - 33593881 - 33653436 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001335.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P50417 AK006993.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P50417 AK006993.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P50417 AK006993.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P50417 AK006993.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P50417 AK006993.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P50417 AK006993.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121262 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136810 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10428067 - -
          Affymetrix HT MG-430 PM Array Plate 1432398_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10428066 - -
          Mouse Genome 430 2.0 Array 1447578_PM_at - -
          Mouse Genome 430 2.0 Array 1432398_PM_at - -
          Mouse Genome 430 2.0 Array 1447578_at - -
          Mouse Genome 430 2.0 Array 1432398_at - -
          Affymetrix Mouse Clariom S Array PSR1500010334.mm.1 TC1500001335.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500010334.mm.1 NR_033608.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500010334.mm.1 NR_033608.1 Refseq
          Mouse Expression Set 430 1447578_at - -
          Mouse Expression Set 430 1432398_PM_at - -
          Mouse Expression Set 430 1447578_PM_at - -
          Mouse Expression Set 430 1432398_at - -
          Affymetrix HT Mouse Genome 430B Array 1432398_at - -
          Affymetrix HT Mouse Genome 430B Array 1447578_at - -
          Affymetrix HT Mouse Genome 430B Array 1432398_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1447578_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 805060 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2706248 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2706248 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2706248 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0