Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04410156_m1
          See other RUNX2 GE Assays ›
          Gene Symbol:
          Runx2
          Gene Name:
          runt related transcription factor 2
          Gene Aliases:
          AML3, Cbf, Cbfa-1, Cbfa1, LS3, Osf2, PEBP2aA, Pebp2a1, Pebpa2a
          Chromosome Location:
          Chr.17: 44495986 - 44814831 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04410156_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Runx2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_073392.1 2 - 3 427 81
          GenBank mRNA AF053954.1 - 2 - 3 257 81
          AF053955.1 - 1 - 2 91 81
          DQ458792.1 - 2 - 3 428 81

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream suppressor of Ty 3 Supt3 2310066G22Rik,
          AI315192,
          SPT3,
          SPT3L,
          Supt3h
          Anti-sense Chr. 17 - 44777171 - 45119294 on NCBI Build GRCm38
          Downstream RIKEN cDNA 4930564C03 gene 4930564C03Rik 65.4kb Sense Chr. 17 - 44879704 - 44880885 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002138.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          osteoblast differentiation
          endochondral ossification
          osteoblast fate commitment
          chondrocyte differentiation
          chondrocyte development
          osteoblast development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription by RNA polymerase II
          positive regulation of cell population proliferation
          positive regulation of gene expression
          hemopoiesis
          neuron differentiation
          T cell differentiation
          regulation of ossification
          BMP signaling pathway
          positive regulation of chondrocyte differentiation
          embryonic forelimb morphogenesis
          regulation of fibroblast growth factor receptor signaling pathway
          odontogenesis of dentin-containing tooth
          regulation of odontogenesis of dentin-containing tooth
          regulation of osteoblast differentiation
          positive regulation of osteoblast differentiation
          positive regulation of ossification
          negative regulation of smoothened signaling pathway
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          cell maturation
          embryonic cranial skeleton morphogenesis
          skeletal system morphogenesis
          stem cell differentiation
          cellular response to BMP stimulus
          positive regulation of transcription from RNA polymerase II promoter involved in cellular response to chemical stimulus
          transcription cis-regulatory region binding
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          DNA binding
          chromatin binding
          DNA-binding transcription factor activity
          protein binding
          ATP binding
          transcription factor binding
          protein domain specific binding
          histone deacetylase binding
          bHLH transcription factor binding
          DNA-binding transcription factor binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700020027.mm.1 TC1700002138.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700020027.mm.1 NR_073392.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020025.mm.1 TC1700002138.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700020025.mm.1 NM_001146038.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020025.mm.1 NM_001271627.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020025.mm.1 NM_009820.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020025.mm.1 NR_073392.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020009.mm.1 TC1700002138.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700020009.mm.1 NM_001145920.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020009.mm.1 NM_001146038.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020009.mm.1 NM_001271627.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020009.mm.1 NM_001271630.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020009.mm.1 NM_001271631.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020009.mm.1 NM_009820.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020009.mm.1 NR_073392.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020009.mm.1 NR_073425.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020019.mm.1 TC1700002138.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700020019.mm.1 NM_001146038.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020019.mm.1 NM_001271627.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020019.mm.1 NM_009820.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020019.mm.1 NR_073392.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020026.mm.1 TC1700002138.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700020026.mm.1 NM_001146038.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020026.mm.1 NM_001271627.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020026.mm.1 NM_009820.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020026.mm.1 NR_073392.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020020.mm.1 TC1700002138.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700020020.mm.1 NM_001146038.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020020.mm.1 NM_001271627.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020020.mm.1 NM_009820.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020020.mm.1 NR_073392.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020022.mm.1 TC1700002138.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700020022.mm.1 NM_001146038.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020022.mm.1 NM_001271627.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020022.mm.1 NM_009820.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020022.mm.1 NR_073392.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 507825 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0