Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04410470_g1
          See other 9330162012RIK GE Assays ›
          Gene Symbol:
          9330162012Rik
          Gene Name:
          cDNA RIKEN 9330162012 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 24937401 - 24939049 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04410470_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9330162012Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_102323.1 1 - 2 205 89
          GenBank mRNA AK034200.1 - 88 89
          AK080139.1 - 1 - 2 182 89
          BC094927.1 - 1 - 2 195 89
          BF465915.1 - 1 - 2 205 89
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BE981625.1 - 1 - 2 204 Chr. 13 - 24937402 - 24938114 [+] on Build GRCm38 89
          GenBankmRNA BB637347.1 - 1 - 2 179 Chr. 13 - 24937427 - 24938212 [+] on Build GRCm38 89
          GenBankmRNA BY229838.1 - 1 - 2 108 Chr. 13 - 24937499 - 24938062 [+] on Build GRCm38 89
          GenBankmRNA CF581768.1 - 1 - 2 238 Chr. 13 - 24937402 - 24938379 [+] on Build GRCm38 89
          GenBankmRNA BB850364.1 - 1 - 2 108 Chr. 13 - 24937499 - 24938097 [+] on Build GRCm38 89
          GenBankmRNA CV674752.1 - 1 - 2 195 Chr. 13 - 24937411 - 24938163 [+] on Build GRCm38 89
          GenBankmRNA BF465914.1 - 1 - 2 205 Chr. 13 - 24937401 - 24938106 [+] on Build GRCm38 89
          GenBankmRNA BY256391.1 - 1 - 2 106 Chr. 13 - 24937500 - 24938121 [+] on Build GRCm38 89
          GenBankmRNA BB626168.1 - 1 - 2 88 Chr. 13 - 24937519 - 24938340 [+] on Build GRCm38 89

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream aldhehyde dehydrogenase family 5,
          subfamily A1
          Aldh5a1 6330403E24Rik,
          Ahd-1,
          Ahd1,
          D630032B01Rik,
          OTTMUSG00000000613,
          SSADH,
          Ssdh1
          Anti-sense Chr. 13 - 24907579 - 24937661 on NCBI Build GRCm38
          Downstream glycosylphosphatidylinositol specific phospholipase D1 Gpld1 6330541J12Rik,
          AW546131
          4.1kb Sense Chr. 13 - 24943152 - 24991936 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0