Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400011344.mm.1 |
TC0400001482.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400011344.mm.1 |
NM_001285885.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011344.mm.1 |
NM_001285887.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011344.mm.1 |
NM_001285888.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011344.mm.1 |
NM_177462.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011344.mm.1 |
NR_104366.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011342.mm.1 |
TC0400001482.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400011342.mm.1 |
NM_001285885.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011342.mm.1 |
NM_001285887.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011342.mm.1 |
NM_001285888.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011342.mm.1 |
NM_177462.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011342.mm.1 |
NR_104366.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011343.mm.1 |
TC0400001482.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400011343.mm.1 |
NM_001285885.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011343.mm.1 |
NM_001285887.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011343.mm.1 |
NM_001285888.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011343.mm.1 |
NM_177462.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011343.mm.1 |
NR_104366.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011345.mm.1 |
TC0400001482.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400011345.mm.1 |
NM_001285885.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011345.mm.1 |
NM_001285887.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011345.mm.1 |
NM_001285888.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011345.mm.1 |
NM_177462.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011345.mm.1 |
NR_104366.1 |
Refseq |