Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04410995_m1
          See other ESR2 GE Assays ›
          Gene Symbol:
          Esr2
          Gene Name:
          estrogen receptor 2 (beta)
          Gene Aliases:
          ER[b], ERbeta, Estrb
          Chromosome Location:
          Chr.12: 76120419 - 76177259 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04410995_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Esr2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_104386.1 5 - 6 1357 92
          GenBank mRNA AK054301.1 - 5 - 6 1358 92

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream spectrin repeat containing,
          nuclear envelope 2
          Syne2 6820443O06Rik,
          AW546258,
          D12Ertd777e,
          NUA,
          Nesp2g,
          mKIAA1011
          -22.9kb Anti-sense Chr. 12 - 75818318 - 76110928 on NCBI Build GRCm38
          Downstream testis expressed gene 21 Tex21 4931406F04Rik,
          4931412D23Rik,
          4931421K24Rik,
          tsec-2
          53.1kb Sense Chr. 12 - 76198687 - 76246758 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          ovarian follicle development
          behavioral fear response
          neuron migration
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription by RNA polymerase II
          brain development
          learning or memory
          negative regulation of cell population proliferation
          hormone-mediated apoptotic signaling pathway
          intracellular steroid hormone receptor signaling pathway
          intracellular estrogen receptor signaling pathway
          response to testosterone
          regulation of cell population proliferation
          positive regulation of apoptotic process
          steroid hormone mediated signaling pathway
          regulation of neuron apoptotic process
          estrous cycle
          positive regulation of epidermal growth factor receptor signaling pathway
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          negative regulation of behavior
          negative regulation of smooth muscle cell proliferation
          negative regulation of epithelial cell proliferation
          positive regulation of DNA-binding transcription factor activity
          Sertoli cell proliferation
          uterus development
          vagina development
          negative regulation of cell death
          prostate gland epithelium morphogenesis
          epithelial cell maturation involved in prostate gland development
          negative regulation of androgen receptor signaling pathway
          positive regulation of ERK1 and ERK2 cascade
          negative regulation of neuron death
          negative regulation of feeding behavior
          negative regulation of reactive oxygen species metabolic process
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          core promoter sequence-specific DNA binding
          DNA binding
          DNA-binding transcription factor activity
          steroid hormone receptor activity
          nuclear receptor activity
          steroid binding
          protein binding
          zinc ion binding
          lipid binding
          enzyme binding
          estrogen receptor activity
          estrogen response element binding
          hormone binding
          peroxisome proliferator activated receptor binding
          sequence-specific DNA binding
          metal ion binding
          estradiol binding
          steroid hormone binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0