Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04411040_m1
          See other HNRNPA1 GE Assays ›
          Gene Symbol:
          Hnrnpa1
          Gene Name:
          heterogeneous nuclear ribonucleoprotein A1
          Gene Aliases:
          HDP-1, Hdp, Hnrpa1, hnRNP A1, hnrnp-A1
          Chromosome Location:
          Chr.15: 103240397 - 103246698 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04411040_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hnrnpa1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_104427.1 9 - 10 1095 65
          GenBank mRNA AK007802.1 - 9 - 10 1041 65
          AK136113.1 - 9 - 10 1065 65
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AW541254.1 - 2 - 3 Chr. 15 - 103242437 - 103246690 [-] on Build GRCm38 65
          GenBankmRNA BM237110.2 - 2 - 3 Chr. 15 - 103243064 - 103246690 [-] on Build GRCm38 65
          GenBankmRNA AA815843.1 - 2 - 3 180 Chr. 15 - 103243672 - 103246691 [+] on Build GRCm38 65
          GenBankmRNA BI788547.1 - 2 - 3 Chr. 15 - 103242451 - 103246671 [-] on Build GRCm38 65
          GenBankmRNA AW550749.2 - 2 - 3 Chr. 15 - 103242697 - 103246690 [-] on Build GRCm38 65
          GenBankmRNA BY754924.1 - 5 - 6 477 Chr. 15 - 103242217 - 103246683 [+] on Build GRCm38 65
          GenBankmRNA AV449278.1 - 2 - 3 88 Chr. 15 - 103242702 - 103246683 [+] on Build GRCm38 65
          GenBankmRNA BB698540.1 - 3 - 4 273 Chr. 15 - 103242672 - 103246689 [+] on Build GRCm38 65
          GenBankmRNA BY470915.1 - 3 - 4 294 Chr. 15 - 103242650 - 103246686 [+] on Build GRCm38 65
          GenBankmRNA CX567652.1 - 4 - 5 387 Chr. 15 - 103242395 - 103246692 [+] on Build GRCm38 65
          GenBankmRNA BM222134.2 - 2 - 3 Chr. 15 - 103242417 - 103246690 [-] on Build GRCm38 65
          GenBankmRNA BP772550.1 - 2 - 3 Chr. 15 - 103242639 - 103246693 [-] on Build GRCm38 65
          GenBankmRNA BQ266428.1 - 2 - 3 Chr. 15 - 103242385 - 103246692 [-] on Build GRCm38 65
          GenBankmRNA AV449224.1 - 2 - 3 89 Chr. 15 - 103242701 - 103246692 [+] on Build GRCm38 65
          GenBankmRNA BY562160.1 - 2 - 3 194 Chr. 15 - 103243660 - 103246693 [+] on Build GRCm38 65
          GenBankmRNA BF320908.1 - 2 - 3 Chr. 15 - 103242241 - 103246692 [-] on Build GRCm38 65
          GenBankmRNA AW046975.1 - 2 - 3 Chr. 15 - 103243698 - 103246693 [-] on Build GRCm38 65
          GenBankmRNA AI314178.1 - Chr. - - [-] on Build 65
          GenBankmRNA BY467940.1 - 4 - 5 335 Chr. 15 - 103242437 - 103246686 [+] on Build GRCm38 65
          GenBankmRNA BY656129.1 - 2 - 3 157 Chr. 15 - 103243695 - 103246696 [+] on Build GRCm38 65
          GenBankmRNA BM241356.2 - 2 - 3 Chr. 15 - 103243722 - 103246693 [-] on Build GRCm38 65
          GenBankmRNA AU019235.1 - 2 - 3 Chr. 15 - 103243631 - 103246690 [-] on Build GRCm38 65
          GenBankmRNA BF140165.1 - 255 Chr. - - [-] on Build 65
          GenBankmRNA AA672806.1 - 2 - 3 Chr. 15 - 103242643 - 103246668 [-] on Build GRCm38 65
          GenBankmRNA AU023093.1 - 2 - 3 Chr. 15 - 103243631 - 103246689 [-] on Build GRCm38 65
          GenBankmRNA BP771005.1 - 2 - 3 Chr. 15 - 103242650 - 103246692 [-] on Build GRCm38 65
          GenBankmRNA BF466706.1 - 2 - 3 244 Chr. 15 - 103242921 - 103246626 [+] on Build GRCm38 65
          GenBankmRNA CJ208873.1 - 2 - 3 193 Chr. 15 - 103243659 - 103246690 [+] on Build GRCm38 65
          GenBankmRNA AA185400.1 - 2 - 3 164 Chr. 15 - 103243701 - 103246692 [+] on Build GRCm38 65

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream chromobox 5 Cbx5 2610029O15Rik,
          C75991,
          HP1,
          Hp1a,
          Hp1alpha
          -4.1kb Anti-sense Chr. 15 - 103191546 - 103239904 on NCBI Build GRCm38
          Downstream nuclear factor,
          erythroid derived 2
          Nfe2 NF-E2,
          NF-E2/P45,
          p45,
          p45NFE2,
          p45nf-e2
          1.8kb Anti-sense Chr. 15 - 103248212 - 103258403 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001127.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          alternative mRNA splicing, via spliceosome
          mRNA processing
          transport
          RNA splicing
          mRNA transport
          nucleotide binding
          nucleic acid binding
          RNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1500009058.mm.1 TC1500001127.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500009058.mm.1 NM_001039129.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1500009058.mm.1 NM_010447.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1500009058.mm.1 NR_002885.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500009058.mm.1 NR_104427.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500009066.mm.1 TC1500001127.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500009066.mm.1 NR_104427.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500009055.mm.1 TC1500001127.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500009055.mm.1 NM_001039129.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1500009055.mm.1 NM_010447.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1500009055.mm.1 NR_002885.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500009055.mm.1 NR_104427.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500009054.mm.1 TC1500001127.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500009054.mm.1 NM_001039129.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1500009054.mm.1 NM_010447.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1500009054.mm.1 NR_002885.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500009054.mm.1 NR_104427.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500009057.mm.1 TC1500001127.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500009057.mm.1 NM_001039129.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1500009057.mm.1 NM_010447.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1500009057.mm.1 NR_002885.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500009057.mm.1 NR_104427.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0