Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04412063_m1
          See other TSPAN5 GE Assays ›
          Gene Symbol:
          Tspan5
          Gene Name:
          tetraspanin 5
          Gene Aliases:
          2810455A09Rik, 4930505M03Rik, NET-4, Tm4sf9
          Chromosome Location:
          Chr.3: 138742208 - 138904433 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04412063_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tspan5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_121195.1 4 - 5 828 114
          GenBank mRNA BC046784.1 - 4 - 5 731 6416107 114
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CF587228.1 - 4 - 5 560 Chr. 3 - 138778200 - 138902494 [+] on Build GRCm38 114
          GenBankmRNA W29958.1 - 112 Chr. - - [-] on Build 114
          GenBankmRNA BU708227.1 - 4 - 5 731 Chr. 3 - 138742305 - 138898148 [+] on Build GRCm38 114
          GenBankmRNA AI391108.1 - 1 - 2 112 Chr. 3 - 138894413 - 138902480 [+] on Build GRCm38 114
          GenBankmRNA BM934469.1 - 4 - 5 Chr. 3 - 138894348 - 138902621 [-] on Build GRCm38 114

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 4930425O10 gene 4930425O10Rik -167.4kb Anti-sense Chr. 3 - 138711535 - 138726984 on NCBI Build GRCm38
          Downstream RAP1,
          GTP-GDP dissociation stimulator 1
          Rap1gds1 BC011279,
          GDS1
          29.0kb Anti-sense Chr. 3 - 138925897 - 139075201 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300001423.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of Notch signaling pathway
          protein maturation
          protein localization to plasma membrane
          enzyme binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300011490.mm.1 TC0300001423.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300011490.mm.1 NM_019571.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0300011492.mm.1 TC0300001423.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300011492.mm.1 NM_019571.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0300011479.mm.1 TC0300001423.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300011479.mm.1 NM_019571.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0300011491.mm.1 TC0300001423.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300011491.mm.1 NM_019571.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0300011499.mm.1 TC0300001423.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300011499.mm.1 NM_019571.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0300011500.mm.1 TC0300001423.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300011500.mm.1 NM_019571.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0300011501.mm.1 TC0300001423.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300011501.mm.1 NM_019571.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0300011497.mm.1 TC0300001423.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300011497.mm.1 NM_019571.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0300011498.mm.1 TC0300001423.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300011498.mm.1 NM_019571.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0300011502.mm.1 TC0300001423.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300011502.mm.1 NM_019571.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0300011480.mm.1 TC0300001423.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300011480.mm.1 NM_019571.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0300011496.mm.1 TC0300001423.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300011496.mm.1 NM_019571.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0300011489.mm.1 TC0300001423.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300011489.mm.1 NM_019571.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0