Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem161a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293797.1 | NP_001280726.1 | 9 - 10 | 1017 | 68 | |
NM_145597.4 | NP_663572.1 | 10 - 11 | 1121 | 68 | ||
NR_122028.1 | 9 - 10 | 1026 | 68 | |||
GenBank mRNA | AK032711.1 | - | 6 - 7 | 637 | 68 | |
AK049979.1 | - | 10 - 11 | 1120 | 68 | ||
AK082758.1 | - | 9 - 10 | 1022 | 68 | ||
AK082916.1 | - | 9 - 10 | 1018 | 68 | ||
AK088569.1 | - | 9 - 10 | 1007 | 68 | ||
AK147962.1 | - | 10 - 11 | 1114 | 68 | ||
AK167517.1 | - | 4 - 5 | 424 | 68 | ||
BC021367.1 | - | 10 - 11 | 1121 | 5030434 | 68 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | myocyte enhancer factor 2B | Mef2b | AI451606 | -13.7kb | Sense | Chr. 8 - 70152778 - 70167488 on NCBI Build GRCm38 |
Downstream |
solute carrier family 25, member 42 |
Slc25a42 | 2900084M01Rik | 2.8kb | Anti-sense | Chr. 8 - 70184340 - 70212281 on NCBI Build GRCm38 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |