Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04934726_m1
          See other BRIP1OS GE Assays ›
          Gene Symbol:
          Brip1os
          Gene Name:
          BRCA1 interacting protein C-terminal helicase 1, opposite strand
          Gene Aliases:
          4632419I22Rik
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04934726_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Brip1os
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK014588.1 - 1 - 2 166 80
          AK052488.1 - 1 - 2 145 80
          AK078546.1 - 1 - 2 178 80
          AK084670.1 - 1 - 2 172 80
          AK132401.1 - 1 - 2 178 80
          BC067002.1 - 1 - 2 187 5716200 80
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB861274.1 - 1 - 2 137 Chr. 11 - 86201414 - 86207266 [+] on Build GRCm38 80
          GenBankmRNA CK788024.1 - 1 - 2 172 Chr. 11 - 86201375 - 86207506 [+] on Build GRCm38 80
          GenBankmRNA AW012521.1 - 1 - 2 165 Chr. 11 - 86201384 - 86207267 [+] on Build GRCm38 80
          GenBankmRNA BU152056.1 - 1 - 2 190 Chr. 11 - 86201393 - 86207663 [+] on Build GRCm38 80
          GenBankmRNA CN666597.1 - 1 - 2 144 Chr. 11 - 86201403 - 86207346 [+] on Build GRCm38 80
          GenBankmRNA CF902967.1 - 1 - 2 168 Chr. 11 - 86201378 - 86207385 [+] on Build GRCm38 80
          GenBankmRNA CJ175467.1 - 1 - 2 206 Chr. 11 - 86201342 - 86207198 [+] on Build GRCm38 80
          GenBankmRNA CA321169.1 - 1 - 2 83 Chr. 11 - 86201464 - 86207566 [+] on Build GRCm38 80
          GenBankmRNA CJ172766.1 - 1 - 2 206 Chr. 11 - 86201342 - 86207173 [+] on Build GRCm38 80
          GenBankmRNA BY714035.1 - 1 - 2 166 Chr. 11 - 86201383 - 86284214 [+] on Build GRCm38 80
          GenBankmRNA BY141999.1 - 1 - 2 177 Chr. 11 - 86201370 - 86207087 [+] on Build GRCm38 80
          GenBankmRNA CJ175122.1 - 1 - 2 206 Chr. 11 - 86201342 - 86207141 [+] on Build GRCm38 80
          GenBankmRNA CF730884.1 - 1 - 2 156 Chr. 11 - 86201391 - 86207411 [+] on Build GRCm38 80
          GenBankmRNA BB623676.1 - 1 - 2 172 Chr. 11 - 86201375 - 86207427 [+] on Build GRCm38 80
          GenBankmRNA BY778546.1 - 1 - 2 171 Chr. 11 - 86201377 - 86207097 [+] on Build GRCm38 80
          GenBankmRNA BY135871.1 - 1 - 2 177 Chr. 11 - 86201370 - 86207145 [+] on Build GRCm38 80
          GenBankmRNA BY776220.1 - 1 - 2 101 Chr. 11 - 86201447 - 86207207 [+] on Build GRCm38 80
          GenBankmRNA BY232584.1 - 1 - 2 177 Chr. 11 - 86201370 - 86207126 [+] on Build GRCm38 80
          GenBankmRNA BY329189.1 - 1 - 2 172 Chr. 11 - 86201375 - 86207100 [+] on Build GRCm38 80
          GenBankmRNA BY782265.1 - 1 - 2 101 Chr. 11 - 86201447 - 86207200 [+] on Build GRCm38 80
          GenBankmRNA BB659241.1 - 1 - 2 170 Chr. 11 - 86201377 - 86207407 [+] on Build GRCm38 80
          GenBankmRNA CF901926.1 - 1 - 2 185 Chr. 11 - 86201362 - 86207422 [+] on Build GRCm38 80
          GenBankmRNA BY136924.1 - 1 - 2 188 Chr. 11 - 86201363 - 86207115 [+] on Build GRCm38 80
          GenBankmRNA BQ571604.1 - 1 - 2 183 Chr. 11 - 86201368 - 86207392 [+] on Build GRCm38 80
          GenBankmRNA CJ178616.1 - 1 - 2 184 Chr. 11 - 86201363 - 86207174 [+] on Build GRCm38 80
          GenBankmRNA BU702503.1 - 1 - 2 131 Chr. 11 - 86201418 - 86207553 [+] on Build GRCm38 80
          GenBankmRNA BB850313.1 - 1 - 2 185 Chr. 11 - 86201364 - 86207171 [+] on Build GRCm38 80
          GenBankmRNA CF725672.1 - 1 - 2 130 Chr. 11 - 86201417 - 86207495 [+] on Build GRCm38 80
          GenBankmRNA CA569102.1 - 1 - 2 76 Chr. 11 - 86201471 - 86207299 [+] on Build GRCm38 80
          GenBankmRNA BB660205.1 - 1 - 2 145 Chr. 11 - 86201403 - 86207455 [+] on Build GRCm38 80
          GenBankmRNA CJ163741.1 - 1 - 2 184 Chr. 11 - 86201364 - 86207201 [+] on Build GRCm38 80
          GenBankmRNA CJ045650.1 - 1 - 2 178 Chr. 11 - 86201370 - 86207421 [+] on Build GRCm38 80
          GenBankmRNA BY143183.1 - 1 - 2 175 Chr. 11 - 86201375 - 86207118 [+] on Build GRCm38 80
          GenBankmRNA BB849135.1 - 1 - 2 184 Chr. 11 - 86201364 - 86207194 [+] on Build GRCm38 79
          GenBankmRNA CF899334.1 - 1 - 2 137 Chr. 11 - 86201413 - 86207448 [+] on Build GRCm38 80
          GenBankmRNA BI683632.1 - 1 - 2 112 Chr. 11 - 86201445 - 86207582 [+] on Build GRCm38 80
          GenBankmRNA CF539445.1 - 1 - 2 153 Chr. 11 - 86201394 - 86207510 [+] on Build GRCm38 80
          GenBankmRNA CJ166692.1 - 1 - 2 206 Chr. 11 - 86201342 - 86207194 [+] on Build GRCm38 80
          GenBankmRNA BY722753.1 - 1 - 2 136 Chr. 11 - 86201411 - 86207406 [+] on Build GRCm38 80
          GenBankmRNA BY138447.1 - 1 - 2 186 Chr. 11 - 86201363 - 86207124 [+] on Build GRCm38 80
          GenBankmRNA CA568104.1 - 1 - 2 178 Chr. 11 - 86201369 - 86207247 [+] on Build GRCm38 80
          GenBankmRNA BY143186.1 - 1 - 2 176 Chr. 11 - 86201375 - 86207117 [+] on Build GRCm38 80
          GenBankmRNA BY299371.1 - 1 - 2 208 Chr. 11 - 86201342 - 86207114 [+] on Build GRCm38 80

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream BRCA1 interacting protein C-terminal helicase 1 Brip1 3110009N10Rik,
          8030460J03Rik,
          Bach1,
          FACJ,
          Fancj,
          OF
          -108.0 Anti-sense Chr. 11 - 86058131 - 86201267 on NCBI Build GRCm38
          Downstream integrator complex subunit 2 Ints2 2810417D08Rik,
          AA408260,
          AI987735,
          mKIAA1287
          3.3kb Anti-sense Chr. 11 - 86210681 - 86257612 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the BC067002.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0