Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm05700833_g1
          See other HSPA8 GE Assays ›
          Gene Symbol:
          Hspa8
          Gene Name:
          heat shock protein 8
          Gene Aliases:
          2410008N15Rik, Hsc70, Hsc71, Hsc73, Hsp73, Hspa10
          Chromosome Location:
          Chr.9: 40801273 - 40805199 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm05700833_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hspa8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031165.4 NP_112442.2 4 - 5 647 84
          GenBank mRNA AK004608.1 - 3 - 4 936 84
          AK035286.1 - 4 - 5 645 84
          AK075935.1 - 4 - 5 644 84
          AK081701.1 - 4 - 5 646 84
          AK098074.1 - 4 - 5 644 84
          AK145416.1 - 4 - 5 567 84
          AK145579.1 - 4 - 5 641 84
          AK146547.1 - 4 - 5 648 84
          AK146708.1 - 4 - 5 642 84
          AK146985.1 - 4 - 5 645 84
          AK147864.1 - 4 - 5 645 84
          AK149954.1 - 4 - 5 642 84
          AK150474.1 - 4 - 5 639 84
          AK150498.1 - 4 - 5 639 84
          AK150701.1 - 4 - 5 649 84
          AK150958.1 - 4 - 5 642 84
          AK151041.1 - 4 - 5 645 84
          AK151065.1 - 4 - 5 645 84
          AK151127.1 - 4 - 5 648 84
          AK151287.1 - 4 - 5 645 84
          AK151435.1 - 4 - 5 645 84
          AK151516.1 - 4 - 5 651 84
          AK151537.1 - 4 - 5 642 84
          AK151741.1 - 4 - 5 645 84
          AK151775.1 - 4 - 5 647 84
          AK151808.1 - 4 - 5 648 84
          AK151865.1 - 4 - 5 645 84
          AK151892.1 - 4 - 5 647 84
          AK151948.1 - 4 - 5 648 84
          AK151997.1 - 4 - 5 645 84
          AK152315.1 - 4 - 5 648 84
          AK152598.1 - 4 - 5 645 84
          AK152697.1 - 4 - 5 645 84
          AK152703.1 - 4 - 5 645 84
          AK152768.1 - 4 - 5 648 84
          AK152803.1 - 4 - 5 648 84
          AK153032.1 - 4 - 5 647 84
          AK153834.1 - 4 - 5 645 84
          AK157831.1 - 4 - 5 645 84
          AK159479.1 - 4 - 5 645 84
          AK163987.1 - 4 - 5 645 84
          AK164000.1 - 4 - 5 645 84
          AK166545.1 - 4 - 5 647 84
          AK166643.1 - 4 - 5 659 84
          AK166721.1 - 4 - 5 642 84
          AK166767.1 - 4 - 5 645 84
          AK166776.1 - 4 - 5 648 84
          AK166808.1 - 4 - 5 645 84
          AK166830.1 - 4 - 5 643 84
          AK166846.1 - 4 - 5 645 84
          AK166861.1 - 4 - 5 648 84
          AK166873.1 - 4 - 5 645 84
          AK166908.1 - 4 - 5 648 84
          AK166910.1 - 4 - 5 645 84
          AK166913.1 - 4 - 5 645 84
          AK166933.1 - 4 - 5 645 84
          AK167043.1 - 4 - 5 647 84
          AK167121.1 - 4 - 5 648 84
          AK167122.1 - 4 - 5 645 84
          AK167134.1 - 4 - 5 643 84
          AK167163.1 - 4 - 5 647 84
          AK167218.1 - 4 - 5 645 84
          AK167229.1 - 4 - 5 646 84
          AK167254.1 - 4 - 5 646 84
          AK167414.1 - 4 - 5 645 84
          AK167845.1 - 4 - 5 648 84
          AK167910.1 - 4 - 5 648 84
          AK168435.1 - 4 - 5 648 84
          AK168492.1 - 4 - 5 647 84
          AK168511.1 - 2 - 3 183 84
          AK168519.1 - 4 - 5 645 84
          AK168542.1 - 4 - 5 647 84
          AK168577.1 - 4 - 5 645 84
          AK168625.1 - 4 - 5 645 84
          AK168654.1 - 4 - 5 645 84
          AK168711.1 - 4 - 5 645 84
          AK168750.1 - 4 - 5 648 84
          AK168776.1 - 4 - 5 648 84
          AK168887.1 - 4 - 5 647 84
          AK168934.1 - 4 - 5 648 84
          AK169093.1 - 4 - 5 645 84
          AK169179.1 - 4 - 5 645 84
          AK169236.1 - 4 - 5 645 84
          AK169293.1 - 4 - 5 659 84
          AK169329.1 - 4 - 5 648 84
          AK169594.1 - 4 - 5 644 84
          AK171434.1 - 4 - 5 646 84
          AK178143.1 - 1 - 2 126 84
          AK184919.1 - 1 - 2 126 84
          AK190509.1 - 1 - 2 126 84
          AK193850.1 - 1 - 2 126 84
          AK195240.1 - 1 - 2 126 84
          AK198438.1 - 1 - 2 126 84
          AK199354.1 - 1 - 2 126 84
          AK199410.1 - 1 - 2 159 84
          AK200094.1 - 1 - 2 84
          AK200309.1 - 1 - 2 126 84
          AK200579.1 - 1 - 2 126 84
          AK201767.1 - 1 - 2 126 84
          AK202859.1 - 1 - 2 126 84
          AK203911.1 - 1 - 2 126 84
          AK204890.1 - 1 - 2 126 84
          AK205187.1 - 1 - 2 126 84
          AK206953.1 - 1 - 2 126 84
          AK210012.1 - 2 - 3 192 84
          AK211519.1 - 1 - 2 126 84
          AK212914.1 - 1 - 2 146 84
          AK214003.1 - 2 - 3 173 84
          AK216159.1 - 1 - 2 126 84
          BC006722.1 - 4 - 5 669 3711399 84
          BC066191.1 - 4 - 5 601 30467052 84
          BC085486.1 - 4 - 5 633 30540914 84
          BC089322.1 - 4 - 5 626 30538640 84
          BC089457.1 - 4 - 5 604 30628505 84
          BC094900.1 - 4 - 5 561 30063483 84
          BC106169.1 - 2 - 3 325 84
          BC106193.1 - 4 - 5 629 6329283 84
          M19141.1 - 4 - 5 631 84
          U27129.1 - 4 - 5 596 84

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hspa8

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          mRNA processing
          protein folding
          RNA splicing
          protein refolding
          negative regulation of transcription, DNA-templated
          ATP metabolic process
          positive regulation of mRNA splicing, via spliceosome
          chaperone mediated protein folding requiring cofactor
          regulation of cell cycle
          chaperone-mediated protein folding
          clathrin coat disassembly
          negative regulation of fibril organization
          nucleotide binding
          G-protein coupled receptor binding
          receptor binding
          protein binding
          ATP binding
          ATPase activity
          enzyme binding
          MHC class II protein complex binding
          heat shock protein binding
          ubiquitin protein ligase binding
          ATPase activity, coupled
          ADP binding
          poly(A) RNA binding
          unfolded protein binding
          C3HC4-type RING finger domain binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline