Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm05700834_g1
          See other HSPA8 GE Assays ›
          Gene Symbol:
          Hspa8
          Gene Name:
          heat shock protein 8
          Gene Aliases:
          2410008N15Rik, Hsc70, Hsc71, Hsc73, Hsp73, Hspa10
          Chromosome Location:
          Chr.9: 40801273 - 40805199 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm05700834_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hspa8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031165.4 NP_112442.2 2 - 3 284 61
          GenBank mRNA AK035286.1 - 2 - 3 282 61
          AK075935.1 - 2 - 3 281 61
          AK081701.1 - 2 - 3 282 61
          AK098074.1 - 2 - 3 281 61
          AK145416.1 - 2 - 3 204 61
          AK145579.1 - 2 - 3 278 61
          AK146547.1 - 2 - 3 285 61
          AK146708.1 - 2 - 3 279 61
          AK146985.1 - 2 - 3 282 61
          AK147864.1 - 2 - 3 282 61
          AK149954.1 - 2 - 3 279 61
          AK150474.1 - 2 - 3 276 61
          AK150498.1 - 2 - 3 276 61
          AK150701.1 - 2 - 3 286 61
          AK150958.1 - 2 - 3 279 61
          AK151041.1 - 2 - 3 282 61
          AK151065.1 - 2 - 3 282 61
          AK151127.1 - 2 - 3 285 61
          AK151287.1 - 2 - 3 282 61
          AK151435.1 - 2 - 3 282 61
          AK151516.1 - 2 - 3 288 61
          AK151537.1 - 2 - 3 279 61
          AK151741.1 - 2 - 3 282 61
          AK151775.1 - 2 - 3 284 61
          AK151808.1 - 2 - 3 285 61
          AK151865.1 - 2 - 3 282 61
          AK151892.1 - 2 - 3 284 61
          AK151948.1 - 2 - 3 285 61
          AK151997.1 - 2 - 3 282 61
          AK152315.1 - 2 - 3 285 61
          AK152598.1 - 2 - 3 282 61
          AK152697.1 - 2 - 3 282 61
          AK152703.1 - 2 - 3 282 61
          AK152768.1 - 2 - 3 285 61
          AK152803.1 - 2 - 3 285 61
          AK153032.1 - 2 - 3 284 61
          AK153834.1 - 2 - 3 282 61
          AK157831.1 - 2 - 3 282 61
          AK159479.1 - 2 - 3 282 61
          AK162775.1 - 2 - 3 282 61
          AK163987.1 - 2 - 3 282 61
          AK164000.1 - 2 - 3 282 61
          AK166643.1 - 2 - 3 296 61
          AK166721.1 - 2 - 3 279 61
          AK166767.1 - 2 - 3 282 61
          AK166776.1 - 2 - 3 285 61
          AK166808.1 - 2 - 3 282 61
          AK166830.1 - 2 - 3 280 61
          AK166846.1 - 2 - 3 282 61
          AK166861.1 - 2 - 3 285 61
          AK166873.1 - 2 - 3 282 61
          AK166908.1 - 2 - 3 285 61
          AK166910.1 - 2 - 3 282 61
          AK166913.1 - 2 - 3 282 61
          AK166933.1 - 2 - 3 282 61
          AK167043.1 - 2 - 3 284 61
          AK167121.1 - 2 - 3 285 61
          AK167122.1 - 2 - 3 282 61
          AK167134.1 - 2 - 3 280 61
          AK167163.1 - 2 - 3 284 61
          AK167218.1 - 2 - 3 282 61
          AK167229.1 - 2 - 3 283 61
          AK167254.1 - 2 - 3 283 61
          AK167414.1 - 2 - 3 282 61
          AK167845.1 - 2 - 3 285 61
          AK167910.1 - 2 - 3 285 61
          AK168435.1 - 2 - 3 285 61
          AK168492.1 - 2 - 3 284 61
          AK168519.1 - 2 - 3 282 61
          AK168542.1 - 2 - 3 284 61
          AK168577.1 - 2 - 3 282 61
          AK168625.1 - 2 - 3 282 61
          AK168654.1 - 2 - 3 282 61
          AK168711.1 - 2 - 3 282 61
          AK168750.1 - 2 - 3 285 61
          AK168776.1 - 2 - 3 285 61
          AK168887.1 - 2 - 3 284 61
          AK168934.1 - 2 - 3 285 61
          AK169093.1 - 2 - 3 282 61
          AK169179.1 - 2 - 3 282 61
          AK169236.1 - 2 - 3 282 61
          AK169293.1 - 2 - 3 296 61
          AK169329.1 - 2 - 3 285 61
          AK169594.1 - 2 - 3 281 61
          AK171434.1 - 2 - 3 283 61
          AK185576.1 - 1 - 2 43 61
          BC006722.1 - 2 - 3 306 3711399 61
          BC066191.1 - 2 - 3 238 30467052 61
          BC085486.1 - 2 - 3 270 30540914 61
          BC089322.1 - 2 - 3 263 30538640 61
          BC089457.1 - 2 - 3 241 30628505 61
          BC094900.1 - 2 - 3 255 30063483 61
          BC106193.1 - 2 - 3 266 6329283 61
          M19141.1 - 2 - 3 268 61
          U27129.1 - 2 - 3 233 61

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hspa8

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          mRNA processing
          protein folding
          RNA splicing
          protein refolding
          negative regulation of transcription, DNA-templated
          ATP metabolic process
          positive regulation of mRNA splicing, via spliceosome
          chaperone cofactor-dependent protein refolding
          regulation of cell cycle
          chaperone-mediated protein folding
          clathrin coat disassembly
          negative regulation of supramolecular fiber organization
          nucleotide binding
          G protein-coupled receptor binding
          RNA binding
          signaling receptor binding
          protein binding
          ATP binding
          ATP hydrolysis activity
          enzyme binding
          MHC class II protein complex binding
          heat shock protein binding
          ubiquitin protein ligase binding
          ADP binding
          unfolded protein binding
          C3HC4-type RING finger domain binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0