Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm05911479_g1
          See other SNORD116 GE Assays ›
          Gene Symbol:
          Snord116 Ipw
          Gene Name:
          small nucleolar RNA, C/D box 116
          imprinted gene in the Prader-Willi syndrome region
          Gene Aliases:
          MBII-85, Pwcr1
          -
          Chromosome Location:
          Chr.7: 59675990 - 59676079 on Build GRCm38
          Chromosome Location:
          Chr.7: 59619158 - 59678878 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm05911479_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Snord116
          Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF241256.1 - LIT1355
          PWCR1
          2420 95
          Gene Symbol: Ipw
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK045353.1 - 1 95

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10564168 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564162 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564164 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564196 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564198 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564182 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564202 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564178 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564200 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564166 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564180 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564194 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564160 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564206 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564184 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564188 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564190 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564192 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564170 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564208 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564174 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564172 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564186 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564176 - -
          Affymetrix HT MG-430 PM Array Plate 1427598_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10564185 - -
          Affymetrix Mouse Gene 1.1 ST Array 10564207 - -
          Affymetrix Mouse Gene 1.1 ST Array 10564187 - -
          Affymetrix Mouse Gene 1.1 ST Array 10564199 - -
          Affymetrix Mouse Gene 1.1 ST Array 10564163 - -
          Affymetrix Mouse Gene 1.1 ST Array 10564177 - -
          Affymetrix Mouse Gene 1.1 ST Array 10564179 - -
          Affymetrix Mouse Gene 1.1 ST Array 10564189 - -
          Affymetrix Mouse Gene 1.1 ST Array 10564201 - -
          Affymetrix Mouse Gene 1.1 ST Array 10564195 - -
          Affymetrix Mouse Gene 1.1 ST Array 10564161 - -
          Affymetrix Mouse Gene 1.1 ST Array 10564175 - -
          Affymetrix Mouse Gene 1.1 ST Array 10564197 - -
          Affymetrix Mouse Gene 1.1 ST Array 10564191 - -
          Affymetrix Mouse Gene 1.1 ST Array 10564169 - -
          Affymetrix Mouse Gene 1.1 ST Array 10564205 - -
          Affymetrix Mouse Gene 1.1 ST Array 10564181 - -
          Affymetrix Mouse Gene 1.1 ST Array 10564167 - -
          Affymetrix Mouse Gene 1.1 ST Array 10564193 - -
          Affymetrix Mouse Gene 1.1 ST Array 10564171 - -
          Affymetrix Mouse Gene 1.1 ST Array 10564183 - -
          Affymetrix Mouse Gene 1.1 ST Array 10564165 - -
          Affymetrix Mouse Gene 1.1 ST Array 10564159 - -
          Affymetrix Mouse Gene 1.1 ST Array 10564173 - -
          Mouse Genome 430 2.0 Array 1427598_PM_at - -
          Mouse Genome 430 2.0 Array 1427598_at - -
          Affymetrix Mouse Clariom S Array PSR0700028557.mm.1 NR_002895.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700028557.mm.1 TC0700003279.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700028545.mm.1 NR_002895.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700028545.mm.1 NR_033779.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700028545.mm.1 TC0700003273.mm.1 MTA TC
          Affymetrix HT Mouse Genome 430A Array 1427598_at - -
          Affymetrix HT Mouse Genome 430A Array 1427598_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0