Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm07293229_m1
          See other ARPP21 GE Assays ›
          Gene Symbol:
          Arpp21
          Gene Name:
          cyclic AMP-regulated phosphoprotein, 21
          Gene Aliases:
          0710001E13Rik, AI853636, ARPP-21, D9Bwg1012e, R3hdm3, Tarpp
          Chromosome Location:
          Chr.9: 112065091 - 112236022 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm07293229_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arpp21
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177615.1 NP_001171086.1 2 - 3 498 70
          NM_001177616.1 NP_001171087.1 3 - 4 505 70
          NM_001177617.1 NP_001171088.1 3 - 4 672 70
          NM_001177618.1 NP_001171089.1 2 - 3 203 70
          NM_001177619.1 NP_001171090.1 3 - 4 870 70
          NM_001177620.1 NP_001171091.1 4 - 5 445 70
          NM_001177623.1 NP_001171094.1 3 - 4 434 70
          NM_028755.3 NP_083031.1 3 - 4 407 70
          NM_033264.2 NP_150289.1 2 - 3 338 70
          GenBank mRNA AF324451.1 - 2 - 3 298 70
          AK002950.2 - 3 - 4 391 70
          AK029467.1 - 3 - 4 871 70
          AK032417.1 - 3 - 4 431 70
          AK032543.1 - 1 - 2 118 70
          AK034376.1 - 1 - 2 51 70
          AK045507.1 - 3 - 4 872 70
          AK050613.1 - 3 - 4 435 70
          AK137974.1 - 3 - 4 673 70
          AK153892.1 - 4 - 5 1153 70
          AK159004.1 - 2 - 3 477 70
          AK160860.1 - 4 - 5 443 70
          BC027107.1 - 3 - 4 395 70
          BC053001.1 - 1 - 2 169 5686446 70
          BC098368.1 - 3 - 4 452 70
          BC128376.1 - 2 - 3 219 70
          BY290692.1 - 2 - 3 204 70

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK159004.1, AK160860.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arpp21

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular response to heat
          molecular_function
          nucleic acid binding
          calmodulin binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0