Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100345389 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017339304.1 | XP_017194793.1 | 737 | 66 |
Set Membership: |
![]() ![]() ![]() |