Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rh00993082_g1
          See other SUPT20H GE Assays ›
          Gene Symbol:
          SUPT20H
          Gene Name:
          SPT20 homolog, SAGA complex component
          Gene Aliases:
          FAM48A
          Chromosome Location:
          Chr.17: 15846771 - 15895883 on Build Mmul_10
          Species:
          Rhesus monkey
          Species Specific ID (Flybase ID):
          -
          Assay ID Rh00993082_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SUPT20H
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_015120926.2 XP_014976412.1 2300 81
          XM_015120927.2 XP_014976413.1 2639 81
          XM_015120929.2 XP_014976415.1 2167 81
          XM_015120930.2 XP_014976416.1 2297 81
          XM_015120931.2 XP_014976417.1 2636 81
          XM_015120932.2 XP_014976418.1 2348 81
          XM_015120933.2 XP_014976419.1 2666 81
          XM_015120934.2 XP_014976420.1 2300 81
          XM_015120936.2 XP_014976422.1 2264 81
          XM_015120937.2 XP_014976423.1 2315 81
          XM_015120939.2 XP_014976425.1 2261 81
          XM_015120940.2 XP_014976426.1 2312 81
          XM_015120941.2 XP_014976427.1 2300 81
          XM_015120942.2 XP_014976428.1 2300 81
          XM_015120943.2 XP_014976429.1 2297 81
          XM_015120944.2 XP_014976430.1 2300 81
          XM_015120945.2 XP_014976431.1 2063 81
          XM_015120946.2 XP_014976432.1 2060 81
          XM_015120947.2 XP_014976433.1 2300 81
          XM_015120948.2 XP_014976434.1 2063 81
          XM_015120949.2 XP_014976435.1 2063 81
          XM_015120950.2 XP_014976436.1 2402 81
          XM_015120951.1 XP_014976437.1 2144 81
          XM_015120952.2 XP_014976438.1 2060 81
          XM_015120953.2 XP_014976439.1 2063 81
          XM_028837428.1 XP_028693261.1 2569 81
          XM_028837429.1 XP_028693262.1 2669 81
          XM_028837430.1 XP_028693263.1 2173 81
          XM_028837431.1 XP_028693264.1 2409 81
          XM_028837433.1 XP_028693266.1 2533 81
          XM_028837435.1 XP_028693268.1 2630 81
          XM_028837436.1 XP_028693269.1 2063 81
          GenBank mRNA JU335347.1 - 21 - 22 2006 81
          JU335348.1 - 21 - 22 2006 81
          JU476308.1 - 21 - 22 1964 81
          JU476309.1 - 21 - 22 1964 81
          JV045758.1 - 21 - 22 1981 81
          JV047582.1 - 21 - 22 2027 81
          JV047583.1 - 21 - 22 2027 81
          JV718351.1 - 21 - 22 2033 81

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Macaca mulatta Probe spans exons

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0