Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260610.1 | NP_001247539.1 | 2 - 3 | 176 | 85 | |
XM_015139570.2 | XP_014995056.1 | 626 | 85 | |||
XM_015139571.2 | XP_014995057.1 | 401 | 85 | |||
XM_028848062.1 | XP_028703895.1 | 131 | 85 | |||
GenBank mRNA | JU333190.1 | - | 2 - 3 | 179 | 85 | |
JU472852.1 | - | 2 - 3 | 176 | 85 |
Set Membership: |
![]() ![]() ![]() ![]() |