Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rh03418534_s1
          See other KIR3DL2 GE Assays ›
          Gene Symbol:
          KIR3DL2
          Gene Name:
          killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail, 2
          Gene Aliases:
          *016:01, *016:02, KIR-3DL, KIR2DL5, KIR3DL06, KIR3DL20, Mamu-KIR
          Chromosome Location:
          Chr.19: 54503377 - 54516676 on Build Mmul_10
          Species:
          Rhesus monkey
          Species Specific ID (Flybase ID):
          -
          Assay ID Rh03418534_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KIR3DL2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001104552.2 NP_001098022.1 4 546 110
          GenBank mRNA AY728184.1 - 4 546 110
          AY728186.1 - 4 546 110
          EU419047.1 - 4 531 110
          EU419048.1 - 4 531 110
          EU419049.1 - 4 531 110
          FJ217807.1 - 4 525 110
          GU112255.1 - 4 540 110
          GU112256.1 - 4 540 110
          GU112264.1 - 4 540 110
          GU112270.1 - 4 540 110
          GU112275.1 - 4 540 110
          GU112289.1 - 4 540 110
          GU112299.1 - 4 540 110
          GU112304.1 - 4 540 110
          GU112308.1 - 4 540 110
          GU112317.1 - 4 540 110
          GU112327.1 - 4 540 110
          GU134802.1 - 4 540 110
          HQ124073.1 - 1 98 110
          LR030464.1 - 4 540 110
          LR030472.1 - 4 504 110
          LR694484.1 - 4 540 110
          LR694485.1 - 4 540 110
          LR694486.1 - 4 540 110
          LR694487.1 - 4 540 110
          LR694488.1 - 4 540 110
          LR694490.1 - 4 540 110
          LR694491.1 - 4 540 110
          LR694492.1 - 4 540 110
          LR694493.1 - 4 540 110
          LR694496.1 - 4 540 110
          LR694497.1 - 4 540 110
          LR694498.1 - 4 540 110
          LR694499.1 - 4 540 110
          LR694500.1 - 4 540 110
          LR694501.1 - 4 540 110
          LR694502.1 - 4 540 110
          LR694503.1 - 4 540 110
          LR694504.1 - 4 540 110
          LR694505.1 - 4 540 110
          LR694506.1 - 4 540 110
          LR694508.1 - 4 540 110
          LR694509.1 - 4 540 110
          LR694510.1 - 4 540 110
          LR694511.1 - 4 540 110
          LR694512.1 - 4 540 110
          LR694513.1 - 4 540 110
          LR694514.1 - 4 540 110
          LR694515.1 - 4 540 110
          LR694516.1 - 4 529 110
          LS997667.1 - 4 540 110
          LS997668.1 - 4 540 110
          LS997669.1 - 4 540 110
          LS997670.1 - 4 540 110
          LS997671.1 - 4 540 110
          LS997672.1 - 4 540 110
          LT906601.1 - 4 540 110
          LT906602.1 - 4 540 110
          LT906603.1 - 4 540 110
          LT906604.1 - 4 540 110
          LT906605.1 - 4 540 110
          LT906606.1 - 4 540 110
          LT906607.1 - 4 540 110
          LT963638.1 - 4 540 110

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Macaca mulatta Both primers and probe map within a single exon

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0