Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rh04345502_m1
See other TFDP2 GE Assays ›
Gene Symbol:
TFDP2
Gene Name:
transcription factor Dp-2
Gene Aliases:
-
Chromosome Location:
Chr.2: 44070437 - 44273083 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID
Rh04345502_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: TFDP2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001261774.1
NP_001248703.1
11 - 12
1389
85
XM_015131927.2
XP_014987413.1
1536
85
XM_015131929.2
XP_014987415.1
1188
85
XM_015131930.2
XP_014987416.1
1249
85
XM_015131931.2
XP_014987417.1
1243
85
XM_015131932.2
XP_014987418.1
1552
85
XM_015131933.2
XP_014987419.1
1142
85
XM_015131934.2
XP_014987420.1
1469
85
XM_015131935.2
XP_014987421.1
1134
85
XM_015131938.2
XP_014987424.1
1411
85
XM_015131939.2
XP_014987425.1
936
85
XM_015131940.2
XP_014987426.1
1363
85
XM_015131941.2
XP_014987427.1
971
85
XM_015131942.2
XP_014987428.1
2649
85
XM_028843366.1
XP_028699199.1
1270
85
XM_028843367.1
XP_028699200.1
1467
85
XM_028843368.1
XP_028699201.1
1398
85
XM_028843369.1
XP_028699202.1
1665
85
XM_028843370.1
XP_028699203.1
1568
85
GenBank mRNA
JU319918.1
-
11 - 12
1236
85
JU319919.1
-
11 - 12
1236
85
JU319920.1
-
11 - 12
1236
85
JU332132.1
-
11 - 12
1236
85
JU472181.1
-
11 - 12
1389
85
JU472182.1
-
11 - 12
1389
85
JU472183.1
-
11 - 12
1389
85
JU472184.1
-
11 - 12
1389
85
JU472185.1
-
11 - 12
1389
85
JU472186.1
-
11 - 12
1389
85
JU472187.1
-
11 - 12
1389
85
JU472188.1
-
11 - 12
1389
85
JU476991.1
-
10 - 11
1266
85
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
Recommended Assay
Macaca mulatta
Probe spans exons
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0