Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rh04959655_s1
          See other CCR5 GE Assays ›
          Gene Symbol:
          CCR5
          Gene Name:
          C-C motif chemokine receptor 5
          Gene Aliases:
          EGK_11711
          Chromosome Location:
          Chr.2: 102388544 - 102394326 on Build Mmul_10
          Species:
          Rhesus monkey
          Species Specific ID (Flybase ID):
          -
          Assay ID Rh04959655_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CCR5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001042773.3 NP_001036238.2 1 1215 106
          NM_001309402.1 NP_001296331.1 2 1020 106
          XM_015131082.2 XP_014986568.1 1239 106
          GenBank mRNA DQ499963.1 - 1 994 106
          DQ499964.1 - 1 994 106
          DQ499965.1 - 1 994 106
          DQ499966.1 - 1 994 106
          DQ499967.1 - 1 994 106
          DQ499968.1 - 1 994 106
          DQ902484.1 - 1 1215 106
          DQ902485.1 - 1 1215 106
          DQ902486.1 - 1 1215 106
          DQ902487.1 - 1 1215 106
          DQ902488.1 - 1 1215 106
          DQ902489.1 - 1 1215 106
          DQ902490.1 - 1 1215 106
          DQ902491.1 - 1 1215 106
          DQ902493.1 - 1 1215 106
          DQ902494.1 - 1 1215 106
          DQ902495.1 - 1 1215 106
          DQ902496.1 - 1 1215 106
          DQ902497.1 - 1 1215 106
          DQ902498.1 - 1 1215 106
          DQ902499.1 - 1 1215 106
          DQ902500.1 - 1 1215 106
          DQ902501.1 - 1 1215 106
          DQ902502.1 - 1 1215 106
          DQ902503.1 - 1 1215 106
          DQ902504.1 - 1 1215 106
          DQ902505.1 - 1 1215 106
          DQ902506.1 - 1 1215 106
          DQ902507.1 - 1 1215 106
          DQ902508.1 - 1 1215 106
          DQ902509.1 - 1 1215 106
          DQ902510.1 - 1 1215 106
          DQ902511.1 - 1 1215 106
          DQ902512.1 - 1 1215 106
          DQ902513.1 - 1 1215 106
          DQ902514.1 - 1 1214 106
          DQ902515.1 - 1 1215 106
          DQ902516.1 - 1 1215 106
          DQ902517.1 - 1 1215 106
          DQ902518.1 - 1 1215 106
          DQ902519.1 - 1 1215 106
          DQ902520.1 - 1 1215 106
          DQ902521.1 - 1 1215 106
          DQ902522.1 - 1 1215 106
          DQ902523.1 - 1 1215 106
          DQ902524.1 - 1 1215 106
          DQ902525.1 - 1 1215 106
          DQ902526.1 - 1 1215 106
          DQ902527.1 - 1 1215 106
          DQ902528.1 - 1 1215 106
          DQ902529.1 - 1 1215 106
          DQ902530.1 - 1 1215 106
          DQ902531.1 - 1 1215 106
          DQ902532.1 - 1 1215 106
          DQ902533.1 - 1 1215 106
          DQ902534.1 - 1 1215 106
          DQ902535.1 - 1 1215 106
          DQ902536.1 - 1 1215 106
          DQ902537.1 - 1 1215 106
          DQ902538.1 - 1 1215 106
          DQ902539.1 - 1 1215 106
          DQ902540.1 - 1 1215 106
          DQ902541.1 - 1 1215 106
          DQ902542.1 - 1 1215 106
          DQ902543.1 - 1 1215 106
          U73739.1 - 1 983 106
          U96762.1 - 1 983 106

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Macaca mulatta Both primers and probe map within a single exon

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0