Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00450822_m1
          See other PFN2 GE Assays ›
          Gene Symbol:
          Pfn2 LOC100909840
          Gene Name:
          profilin 2
          profilin-2-like
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.2: 147953863 - 147959692 on Build Rnor_6.0
          Chromosome Location:
          Chr.2: 148044717 - 148050464 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn00450822_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pfn2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030873.1 NP_110500.1 327 104
          XM_006232390.2 XP_006232452.1 2 - 3 452 104
          GenBank mRNA AF228736.1 - 327 104
          AF228737.1 - 327 104
          AY004289.1 - 327 104
          BC087013.1 - 330 104
          Gene Symbol: LOC100909840
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003749282.2 XP_003749330.1 2 - 3 368 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003743.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pfn2
          Click here to find siRNAs for Gene LOC100909840

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of epithelial cell migration
          actin cytoskeleton organization
          regulation of actin filament polymerization
          negative regulation of actin filament polymerization
          positive regulation of actin filament polymerization
          positive regulation of actin filament bundle assembly
          positive regulation of ATPase activity
          positive regulation of peptidyl-serine phosphorylation
          positive regulation of stress fiber assembly
          negative regulation of ruffle assembly
          regulation of synaptic vesicle exocytosis
          adenyl-nucleotide exchange factor activity
          actin binding
          actin monomer binding
          phosphatidylinositol-4,5-bisphosphate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P581863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872419 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118985 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P581863 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P581863 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P581863 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P581863 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76056 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10823308 - -
          Affymetrix Rat Gene 1.0 ST Array 10823306 - -
          Affymetrix Rat Gene 1.0 ST Array 10823307 - -
          Affymetrix Rat Gene 1.0 ST Array 10823304 - -
          Affymetrix Rat Gene 1.0 ST Array 10823305 - -
          Affymetrix Rat Gene 1.1 ST Array 10823303 - -
          Rat Genome 230 2.0 Array 1387015_PM_at - -
          Rat Genome 230 2.0 Array 1367970_at - -
          Rat Genome 230 2.0 Array 1387015_at - -
          Rat Genome 230 2.0 Array 1367970_PM_at - -
          Affymetrix HT Rat Focus Array 1387015_at - -
          Affymetrix HT Rat Focus Array 1367970_PM_at - -
          Affymetrix HT Rat Focus Array 1387015_PM_at - -
          Affymetrix HT Rat Focus Array 1367970_at - -
          Affymetrix Rat Clariom S Assay PSR0200025080.rn.1 TC0200003743.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200025079.rn.1 TC0200003743.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200025078.rn.1 TC0200003743.rn.1 RTA TC
          Rat Expression Set 230 1367970_at - -
          Rat Expression Set 230 1367970_PM_at - -
          Rat Expression Set 230 1387015_PM_at - -
          Rat Expression Set 230 1387015_at - -
          Affymetrix Rat Gene 1.0 ST Array 10823303 - -
          Rat Genome U34 Set rc_AA943544_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22220443 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370459 - -
          RatRef-12 Expression BeadChip ILMN_66748 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0