Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00456287_m1
          See other STMN3 GE Assays ›
          Gene Symbol:
          Stmn3
          Gene Name:
          stathmin-like 3
          Gene Aliases:
          Sclip
          Chromosome Location:
          Chr.3: 176808016 - 176816108 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00456287_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stmn3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024346.1 NP_077322.1 4 - 5 509 89
          GenBank mRNA AY004290.1 - 4 - 5 509 89
          FQ211987.1 - 3 - 4 361 89
          FQ213831.1 - 3 - 4 364 89
          FQ214184.1 - 3 - 4 366 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300005050.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Stmn3

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule depolymerization
          regulation of microtubule polymerization or depolymerization
          cytoplasmic microtubule organization
          neuron projection development
          negative regulation of Rac protein signal transduction
          regulation of cytoskeleton organization
          protein binding
          protein C-terminus binding
          tubulin binding
          protein domain specific binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P059775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759792 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130207 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P059775 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P059775 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P059775 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1042234 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700526077 - -
          Agilent Rat cDNA Microarray (G4105A) 700526077 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10852532 - -
          Affymetrix Rat Gene 1.0 ST Array 10852538 - -
          Affymetrix Rat Gene 1.0 ST Array 10852537 - -
          Affymetrix Rat Gene 1.0 ST Array 10852533 - -
          Affymetrix Rat Gene 1.0 ST Array 10852534 - -
          Affymetrix Rat Gene 1.0 ST Array 10852536 - -
          Affymetrix Rat Gene 1.1 ST Array 10852531 - -
          Rat Genome 230 2.0 Array 1368157_PM_at - -
          Rat Genome 230 2.0 Array 1368157_at - -
          Affymetrix HT Rat Focus Array 1368157_at - -
          Affymetrix HT Rat Focus Array 1368157_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300038409.rn.1 TC0300005050.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038418.rn.1 TC0300005050.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038421.rn.1 TC0300005050.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038413.rn.1 TC0300005050.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038412.rn.1 TC0300005050.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038417.rn.1 TC0300005050.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038410.rn.1 TC0300005050.rn.1 RTA TC
          Rat Expression Set 230 1368157_PM_at - -
          Rat Expression Set 230 1368157_at - -
          Affymetrix Rat Gene 1.0 ST Array 10852531 - -
          Rat Genome U34 Set rc_AA899649_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21149196 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355500 - -
          RatRef-12 Expression BeadChip ILMN_55064 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-998b77544-nf2nv:80/100.66.134.75:80.
          git-commit: efd2b0e08ab99238c7874a09efb06372d741ac0a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc