Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108046.2 | NP_001101516.1 | 3 - 4 | 302 | 94 | |
GenBank mRNA | BC166825.1 | - | 3 - 4 | 302 | 94 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |