Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00562004_m1
          See other TACR1 GE Assays ›
          Gene Symbol:
          Tacr1
          Gene Name:
          tachykinin receptor 1
          Gene Aliases:
          Tac1r
          Chromosome Location:
          Chr.4: 113247236 - 113416139 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00562004_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tacr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012667.2 NP_036799.1 1141 69
          GenBank mRNA J05097.1 - 1141 69
          M31477.1 - 2 - 3 598 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001215.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tacr1

          Gene Ontology Categories:

          Function(s) Process(es)

          aggressive behavior
          acute inflammatory response
          positive regulation of leukocyte migration
          angiotensin-mediated drinking behavior
          positive regulation of cytosolic calcium ion concentration
          tachykinin receptor signaling pathway
          neuropeptide signaling pathway
          learning or memory
          long-term memory
          regulation of blood pressure
          associative learning
          response to heat
          response to hormone
          response to ozone
          positive regulation of epithelial cell migration
          response to auditory stimulus
          response to organic cyclic compound
          regulation of smooth muscle cell migration
          sensory perception of pain
          positive regulation of synaptic transmission, cholinergic
          positive regulation of synaptic transmission, GABAergic
          response to estradiol
          response to progesterone
          response to nicotine
          operant conditioning
          positive regulation of renal sodium excretion
          sperm ejaculation
          eating behavior
          positive regulation of vascular permeability
          response to morphine
          response to ethanol
          positive regulation of action potential
          positive regulation of blood pressure
          positive regulation of ossification
          positive regulation of vasoconstriction
          positive regulation of saliva secretion
          positive regulation of hormone secretion
          response to pain
          behavioral response to pain
          regulation of smooth muscle cell proliferation
          positive regulation of lymphocyte proliferation
          positive regulation of epithelial cell proliferation
          positive regulation of stress fiber assembly
          response to electrical stimulus
          smooth muscle contraction involved in micturition
          positive regulation of uterine smooth muscle contraction
          substance P receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P215075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869139 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112305 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119911 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18546 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141900 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57477 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7245 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104592 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117983 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118465 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154692 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108341 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52333 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99081 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14346 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14707 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66338 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176100 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156466 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149895 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5739 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P215075 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P215075 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P215075 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P215075 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11366 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23114 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36221 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48579 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68934 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61134 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93590 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88424 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36466 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67090 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10856526 - -
          Affymetrix Rat Gene 1.0 ST Array 10856531 - -
          Affymetrix Rat Gene 1.0 ST Array 10856528 - -
          Affymetrix Rat Gene 1.0 ST Array 10856527 - -
          Affymetrix Rat Gene 1.0 ST Array 10856529 - -
          Affymetrix Rat Gene 1.0 ST Array 10856530 - -
          Affymetrix Rat Gene 1.0 ST Array 10856532 - -
          Affymetrix Rat Gene 1.1 ST Array 10856525 - -
          Rat Genome 230 2.0 Array 1369199_at - -
          Rat Genome 230 2.0 Array 1387474_at - -
          Rat Genome 230 2.0 Array 1387474_PM_at - -
          Rat Genome 230 2.0 Array 1369199_PM_at - -
          Affymetrix HT Rat Focus Array 1369199_at - -
          Affymetrix HT Rat Focus Array 1387474_PM_at - -
          Affymetrix HT Rat Focus Array 1387474_at - -
          Affymetrix HT Rat Focus Array 1369199_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400008462.rn.1 TC0400001215.rn.1 RTA TC
          Rat Expression Set 230 1369199_PM_at - -
          Rat Expression Set 230 1369199_at - -
          Rat Expression Set 230 1387474_PM_at - -
          Rat Expression Set 230 1387474_at - -
          Affymetrix Rat Gene 1.0 ST Array 10856525 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21063330 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376350 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0