Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00563503_m1
          See other OXTR GE Assays ›
          Gene Symbol:
          Oxtr
          Gene Name:
          oxytocin receptor
          Gene Aliases:
          OTR, OTR1
          Chromosome Location:
          Chr.4: 144401098 - 144417598 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00563503_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Oxtr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012871.2 NP_037003.2 1 - 2 918 60
          XM_006237008.2 XP_006237070.1 2 - 3 2043 60
          XM_008763179.1 XP_008761401.1 2 - 3 2352 60
          XM_008763180.1 XP_008761402.1 2 - 3 2140 60
          XM_008763181.1 XP_008761403.1 2 - 3 2333 60
          XM_008763182.1 XP_008761404.1 2 - 3 1847 60
          GenBank mRNA AY338194.1 - 1 - 2 918 60
          L81169.1 - 1 - 2 948 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003515.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Oxtr

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          suckling behavior
          response to amphetamine
          regulation of systemic arterial blood pressure by vasopressin
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          heart development
          female pregnancy
          lactation
          memory
          positive regulation of norepinephrine secretion
          response to organic cyclic compound
          telencephalon development
          sleep
          positive regulation of synaptic transmission, GABAergic
          response to estradiol
          response to progesterone
          cellular response to hormone stimulus
          response to anoxia
          response to cytokine
          social behavior
          response to cocaine
          response to drug
          maternal behavior
          sperm ejaculation
          eating behavior
          response to peptide hormone
          regulation of digestive system process
          estrous cycle
          positive regulation of blood pressure
          positive regulation of vasoconstriction
          response to steroid hormone
          digestive tract development
          positive regulation of synapse assembly
          positive regulation of synaptic transmission, glutamatergic
          maternal process involved in parturition
          positive regulation of penile erection
          negative regulation of gastric acid secretion
          ERK1 and ERK2 cascade
          positive regulation of uterine smooth muscle contraction
          oxytocin receptor activity
          vasopressin receptor activity
          protein binding
          peptide hormone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P092259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556332 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42320 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152137 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P092259 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P092259 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P092259 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P288043 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99430 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4897 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10864571 - -
          Affymetrix Rat Gene 1.0 ST Array 10864570 - -
          Affymetrix Rat Gene 1.1 ST Array 10864569 - -
          Rat Genome 230 2.0 Array 1387710_PM_at - -
          Rat Genome 230 2.0 Array 1387710_at - -
          Affymetrix HT Rat Focus Array 1387710_PM_at - -
          Affymetrix HT Rat Focus Array 1387710_at - -
          Affymetrix Rat Clariom S Assay PSR0400024657.rn.1 TC0400003515.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400024658.rn.1 TC0400003515.rn.1 RTA TC
          Rat Expression Set 230 1387710_at - -
          Rat Expression Set 230 1387710_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10864569 - -
          Rat Genome U34 Set U15280_at - -
          Rat Genome U34 Set S79241_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21296502 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650850 - -
          RatRef-12 Expression BeadChip ILMN_68235 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0