Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00563902_m1
          See other CASP3 GE Assays ›
          Gene Symbol:
          Casp3
          Gene Name:
          caspase 3
          Gene Aliases:
          Lice
          Chromosome Location:
          Chr.16: 48845011 - 48863243 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00563902_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Casp3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012922.2 NP_037054.1 3 - 4 230 93
          XM_006253130.2 XP_006253192.1 3 - 4 274 93
          GenBank mRNA BC081854.1 - 3 - 4 230 7104685 93
          U34685.1 - 1 - 2 88 93
          U49930.1 - 2 - 3 200 93
          U84410.1 - 2 - 3 204 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600001643.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protease Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Casp3

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          B cell homeostasis
          release of cytochrome c from mitochondria
          proteolysis
          apoptotic process
          cellular response to DNA damage stimulus
          heart development
          sensory perception of sound
          learning or memory
          extrinsic apoptotic signaling pathway via death domain receptors
          intrinsic apoptotic signaling pathway in response to oxidative stress
          response to UV
          response to wounding
          response to glucose
          response to organic substance
          response to metal ion
          response to X-ray
          response to organic cyclic compound
          protein processing
          hippocampus development
          neuron differentiation
          keratinocyte differentiation
          erythrocyte differentiation
          negative regulation of B cell proliferation
          response to cobalt ion
          response to estradiol
          response to lipopolysaccharide
          glial cell apoptotic process
          response to nicotine
          intracellular signal transduction
          wound healing
          response to drug
          response to hydrogen peroxide
          T cell homeostasis
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of catalytic activity
          response to amino acid
          positive regulation of neuron apoptotic process
          cell fate commitment
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          negative regulation of cell cycle
          negative regulation of activated T cell proliferation
          response to antibiotic
          response to glucocorticoid
          neuron apoptotic process
          cellular response to organic substance
          cellular response to organic cyclic compound
          extrinsic apoptotic signaling pathway in absence of ligand
          execution phase of apoptosis
          regulation of apoptotic DNA fragmentation
          protease binding
          aspartic-type endopeptidase activity
          cysteine-type endopeptidase activity
          cyclin-dependent protein serine/threonine kinase inhibitor activity
          death receptor binding
          protein binding
          peptidase activity
          cysteine-type peptidase activity
          phospholipase A2 activator activity
          protein complex binding
          cysteine-type endopeptidase activity involved in apoptotic process
          cysteine-type endopeptidase activity involved in execution phase of apoptosis

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P530813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274049 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161059 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17048 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P530813 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P530813 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P530813 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P496262 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P530813 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23326 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100426 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10791656 - -
          Affymetrix Rat Gene 1.0 ST Array 10791658 - -
          Affymetrix Rat Gene 1.0 ST Array 10791654 - -
          Affymetrix Rat Gene 1.0 ST Array 10791653 - -
          Affymetrix Rat Gene 1.0 ST Array 10791657 - -
          Affymetrix Rat Gene 1.0 ST Array 10791659 - -
          Affymetrix Rat Gene 1.0 ST Array 10791655 - -
          Affymetrix Rat Gene 1.1 ST Array 10791652 - -
          Rat Genome 230 2.0 Array 1390386_at - -
          Rat Genome 230 2.0 Array 1387690_at - -
          Rat Genome 230 2.0 Array 1387690_PM_at - -
          Rat Genome 230 2.0 Array 1390386_PM_at - -
          Affymetrix HT Rat Focus Array 1390386_at - -
          Affymetrix HT Rat Focus Array 1387690_PM_at - -
          Affymetrix HT Rat Focus Array 1387690_at - -
          Affymetrix HT Rat Focus Array 1390386_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600012246.rn.1 TC1600001643.rn.1 RTA TC
          Rat Expression Set 230 1390386_PM_at - -
          Rat Expression Set 230 1387690_PM_at - -
          Rat Expression Set 230 1387690_at - -
          Rat Expression Set 230 1390386_at - -
          Affymetrix Rat Gene 1.0 ST Array 10791652 - -
          Rat Genome U34 Set U49930_g_at - -
          Rat Genome U34 Set U84410_s_at - -
          Rat Genome U34 Set U49930_at - -
          Rat Genome U34 Set rc_AI059604_at - -
          Rat Genome U34 Set U49930_g_at - -
          Rat Genome U34 Set U84410_s_at - -
          Rat Genome U34 Set U49930_at - -
          Rat Genome U34 Set U84410_s_at - -
          Rat Genome U34 Set U49930_g_at - -
          Rat Genome U34 Set U49930_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22257881 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349218 - -
          RatRef-12 Expression BeadChip ILMN_50498 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0