Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00564055_m1
          See other CYP1B1 GE Assays ›
          Gene Symbol:
          Cyp1b1
          Gene Name:
          cytochrome P450, family 1, subfamily b, polypeptide 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 2310988 - 2316722 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00564055_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp1b1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012940.2 NP_037072.1 1 - 2 371 69
          GenBank mRNA U09540.1 - 371 69

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the NM_012940.2, U09540.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 P450 Detect Genomic DNA Rattus norvegicus Rat Estrogen Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyp1b1

          Panther Classification:

          Molecular Function -

          oxidoreductase oxygenase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          trabecular meshwork development
          DNA modification
          cellular aromatic compound metabolic process
          xenobiotic metabolic process
          nitric oxide biosynthetic process
          cell adhesion
          response to nutrient
          steroid metabolic process
          estrogen metabolic process
          negative regulation of cell proliferation
          male gonad development
          intrinsic apoptotic signaling pathway in response to oxidative stress
          toxin metabolic process
          response to toxic substance
          response to organic substance
          positive regulation of vascular endothelial growth factor production
          response to organic cyclic compound
          positive regulation of smooth muscle cell migration
          dibenzo-p-dioxin metabolic process
          arachidonic acid metabolic process
          collagen fibril organization
          adrenal gland development
          negative regulation of cell migration
          negative regulation of NF-kappaB transcription factor activity
          response to follicle-stimulating hormone
          response to estradiol
          negative regulation of cell adhesion mediated by integrin
          benzene-containing compound metabolic process
          retinol metabolic process
          retinal metabolic process
          positive regulation of apoptotic process
          endothelial cell migration
          estrous cycle
          positive regulation of translation
          positive regulation of angiogenesis
          positive regulation of JAK-STAT cascade
          membrane lipid catabolic process
          response to arsenic-containing substance
          blood vessel morphogenesis
          response to steroid hormone
          oxidation-reduction process
          retina vasculature development in camera-type eye
          retinal blood vessel morphogenesis
          ganglion development
          cellular response to hydrogen peroxide
          cellular response to cAMP
          cellular response to tumor necrosis factor
          cellular response to luteinizing hormone stimulus
          cellular response to cortisol stimulus
          cellular response to progesterone stimulus
          cellular response to organic cyclic compound
          response to dexamethasone
          endothelial cell-cell adhesion
          response to indole-3-methanol
          regulation of reactive oxygen species metabolic process
          positive regulation of reactive oxygen species metabolic process
          positive regulation of DNA biosynthetic process
          monooxygenase activity
          iron ion binding
          oxidoreductase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
          heme binding
          aromatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P126969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339710 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75527 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P126969 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P126969 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P126969 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P251228 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P191778 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700183177 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73047 - -
          Agilent Rat cDNA Microarray (G4105A) 700183177 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10887951 - -
          Affymetrix Rat Gene 1.0 ST Array 10887949 - -
          Affymetrix Rat Gene 1.0 ST Array 10882491 - -
          Affymetrix Rat Gene 1.0 ST Array 10887950 - -
          Affymetrix Rat Gene 1.0 ST Array 10887948 - -
          Affymetrix Rat Gene 1.1 ST Array 10887947 - -
          Affymetrix Rat Gene 1.1 ST Array 10882490 - -
          Rat Genome 230 2.0 Array 1368990_at - -
          Rat Genome 230 2.0 Array 1368990_PM_at - -
          Affymetrix HT Rat Focus Array 1368990_PM_at - -
          Affymetrix HT Rat Focus Array 1368990_at - -
          Affymetrix Rat Clariom S Assay PSR0600012389.rn.1 TC0600001745.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600012392.rn.1 TC0600001745.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600012390.rn.1 TC0600001745.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600012397.rn.1 TC0600001745.rn.1 RTA TC
          Rat Expression Set 230 1368990_PM_at - -
          Rat Expression Set 230 1368990_at - -
          Affymetrix Rat Gene 1.0 ST Array 10887947 - -
          Rat Genome U34 Set rc_AI176856_at - -
          Rat Genome U34 Set U09540_at - -
          Rat Genome U34 Set X83867cds_s_at - -
          Rat Genome U34 Set U09540_g_at - -
          Rat Genome U34 Set U09540_at - -
          Rat Genome U34 Set X83867cds_s_at - -
          Rat Genome U34 Set U09540_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20696290 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376635 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline