Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00564292_m1
          See other LGALS4 GE Assays ›
          Gene Symbol:
          Lgals4
          Gene Name:
          lectin, galactoside-binding, soluble, 4
          Gene Aliases:
          L-36, L36LBI
          Chromosome Location:
          Chr.1: 87019808 - 87027720 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00564292_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lgals4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012975.1 NP_037107.1 4 - 5 494 99
          XM_008759113.1 XP_008757335.1 4 - 5 661 99
          GenBank mRNA M73553.1 - 4 - 5 494 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001193.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lgals4

          Gene Ontology Categories:

          Process(es)

          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P109899 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P109900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622849 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106876 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3778 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P109900 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P109899 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P109899 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P109900 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P109899 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P109900 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P491119 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40980 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13947 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10705651 - -
          Affymetrix Rat Gene 1.0 ST Array 10705652 - -
          Affymetrix Rat Gene 1.0 ST Array 10705648 - -
          Affymetrix Rat Gene 1.0 ST Array 10705646 - -
          Affymetrix Rat Gene 1.0 ST Array 10705650 - -
          Affymetrix Rat Gene 1.0 ST Array 10705643 - -
          Affymetrix Rat Gene 1.0 ST Array 10705647 - -
          Affymetrix Rat Gene 1.0 ST Array 10705645 - -
          Affymetrix Rat Gene 1.0 ST Array 10705644 - -
          Affymetrix Rat Gene 1.0 ST Array 10705649 - -
          Affymetrix Rat Gene 1.1 ST Array 10705642 - -
          Rat Genome 230 2.0 Array 1368269_PM_at - -
          Rat Genome 230 2.0 Array 1368269_at - -
          Affymetrix HT Rat Focus Array 1368269_PM_at - -
          Affymetrix HT Rat Focus Array 1368269_at - -
          Affymetrix Rat Clariom S Assay PSR0100009039.rn.1 TC0100001193.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100009038.rn.1 TC0100001193.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100009040.rn.1 TC0100001193.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100009041.rn.1 TC0100001193.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100009042.rn.1 TC0100001193.rn.1 RTA TC
          Rat Expression Set 230 1368269_PM_at - -
          Rat Expression Set 230 1368269_at - -
          Affymetrix Rat Gene 1.0 ST Array 10705642 - -
          Rat Genome U34 Set M73553_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21388624 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364084 - -
          RatRef-12 Expression BeadChip ILMN_70334 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0