Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00566727_m1
          See other ITGB1 GE Assays ›
          Gene Symbol:
          Itgb1
          Gene Name:
          integrin, beta 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 61677512 - 61725537 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00566727_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Itgb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017022.2 NP_058718.2 6 - 7 957 81
          XM_006255824.1 XP_006255886.1 6 - 7 987 81
          GenBank mRNA BC131845.1 - 6 - 7 957 8366766 81
          U12309.1 - 6 - 7 831 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000902.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Inflammation 1700 Inventoried Rat_Inflammation Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Itgb1

          Panther Classification:

          Molecular Function -

          integrin

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          in utero embryonic development
          cell fate specification
          tissue homeostasis
          cell migration involved in sprouting angiogenesis
          cellular calcium ion homeostasis
          cell adhesion
          leukocyte cell-cell adhesion
          cell-matrix adhesion
          calcium-independent cell-matrix adhesion
          transforming growth factor beta receptor signaling pathway
          integrin-mediated signaling pathway
          regulation of G-protein coupled receptor protein signaling pathway
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          germ cell migration
          visual learning
          regulation of collagen catabolic process
          positive regulation of cell-substrate adhesion
          positive regulation of neuron projection development
          response to activity
          cell migration
          formation of radial glial scaffolds
          positive regulation of cell migration
          neuron projection development
          negative regulation of cell projection organization
          cell-substrate adhesion
          response to nutrient levels
          protein transport within lipid bilayer
          cell-cell adhesion mediated by integrin
          heterotypic cell-cell adhesion
          response to gonadotropin
          negative regulation of Rho protein signal transduction
          response to drug
          positive regulation of apoptotic process
          stress fiber assembly
          positive regulation of MAPK cascade
          sarcomere organization
          negative regulation of cell differentiation
          negative regulation of neuron differentiation
          positive regulation of neuron differentiation
          positive regulation of endocytosis
          mesodermal cell differentiation
          axon extension
          cardiac muscle tissue development
          dendrite morphogenesis
          positive regulation of peptidyl-tyrosine phosphorylation
          leukocyte tethering or rolling
          regulation of cell cycle
          cardiac muscle cell differentiation
          maternal process involved in female pregnancy
          bicellular tight junction assembly
          cellular response to mechanical stimulus
          cellular response to vitamin D
          cellular response to ionizing radiation
          response to transforming growth factor beta
          positive regulation of establishment of protein localization to plasma membrane
          negative regulation of anoikis
          glycoprotein binding
          fibronectin binding
          protease binding
          actin binding
          receptor activity
          receptor binding
          integrin binding
          protein binding
          collagen binding
          kinase binding
          protein kinase binding
          protein domain specific binding
          protein complex binding
          peptide binding
          laminin binding
          metal ion binding
          protein heterodimerization activity
          cell adhesion molecule binding
          alpha-actinin binding
          collagen binding involved in cell-matrix adhesion

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P273697 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P507943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949783 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3890 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69148 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174464 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158613 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53202 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P273697 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P507943 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P507943 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P273697 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P507943 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P273697 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P273697 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700294207 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700275551 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12092 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 340 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61159 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31219 - -
          Agilent Rat cDNA Microarray (G4105A) 700275551 - -
          Agilent Rat cDNA Microarray (G4105A) 700294207 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10812032 - -
          Affymetrix Rat Gene 1.0 ST Array 10812035 - -
          Affymetrix Rat Gene 1.0 ST Array 10812029 - -
          Affymetrix Rat Gene 1.0 ST Array 10812037 - -
          Affymetrix Rat Gene 1.0 ST Array 10812027 - -
          Affymetrix Rat Gene 1.0 ST Array 10812023 - -
          Affymetrix Rat Gene 1.0 ST Array 10812022 - -
          Affymetrix Rat Gene 1.0 ST Array 10812036 - -
          Affymetrix Rat Gene 1.0 ST Array 10808983 - -
          Affymetrix Rat Gene 1.0 ST Array 10812038 - -
          Affymetrix Rat Gene 1.0 ST Array 10812028 - -
          Affymetrix Rat Gene 1.0 ST Array 10812033 - -
          Affymetrix Rat Gene 1.0 ST Array 10812034 - -
          Affymetrix Rat Gene 1.0 ST Array 10812031 - -
          Affymetrix Rat Gene 1.0 ST Array 10812026 - -
          Affymetrix Rat Gene 1.0 ST Array 10812039 - -
          Affymetrix Rat Gene 1.0 ST Array 10812030 - -
          Affymetrix Rat Gene 1.1 ST Array 10812021 - -
          Rat Genome 230 2.0 Array 1368819_at - -
          Rat Genome 230 2.0 Array 1368819_PM_at - -
          Rat Genome 230 2.0 Array 1387346_at - -
          Rat Genome 230 2.0 Array 1387346_PM_at - -
          Affymetrix HT Rat Focus Array 1368819_PM_at - -
          Affymetrix HT Rat Focus Array 1387346_PM_at - -
          Affymetrix HT Rat Focus Array 1387346_at - -
          Affymetrix HT Rat Focus Array 1368819_at - -
          Affymetrix Rat Clariom S Assay PSR1900007223.rn.1 TC1900000902.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900007238.rn.1 TC1900000902.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900007233.rn.1 TC1900000902.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900007224.rn.1 TC1900000902.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900007228.rn.1 TC1900000902.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900007230.rn.1 TC1900000902.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900007232.rn.1 TC1900000902.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900007234.rn.1 TC1900000902.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900007225.rn.1 TC1900000902.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900007229.rn.1 TC1900000902.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900007227.rn.1 TC1900000902.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900007231.rn.1 TC1900000902.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900007226.rn.1 TC1900000902.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900007236.rn.1 TC1900000902.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900007235.rn.1 TC1900000902.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900007237.rn.1 TC1900000902.rn.1 RTA TC
          Rat Expression Set 230 1387346_PM_at - -
          Rat Expression Set 230 1368819_PM_at - -
          Rat Expression Set 230 1387346_at - -
          Rat Expression Set 230 1368819_at - -
          Affymetrix Rat Gene 1.0 ST Array 10812021 - -
          Rat Genome U34 Set rc_AI177366_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21590461 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361299 - -
          RatRef-12 Expression BeadChip ILMN_63152 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0