Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00566763_m1
          See other NEFM GE Assays ›
          Gene Symbol:
          Nefm
          Gene Name:
          neurofilament, medium polypeptide
          Gene Aliases:
          Nef3, Nfm
          Chromosome Location:
          Chr.15: 44855307 - 44860647 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00566763_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nefm
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017029.2 NP_058725.1 2 - 3 1236 110
          GenBank mRNA M18628.1 - 2 - 3 1214 110
          Z12152.1 - 2 - 3 1239 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001884.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nefm

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          axo-dendritic transport
          spinal cord development
          axon regeneration
          regulation of axon diameter
          cellular response to oxidative stress
          intermediate filament cytoskeleton organization
          intermediate filament polymerization or depolymerization
          intermediate filament bundle assembly
          neurofilament cytoskeleton organization
          cellular response to estradiol stimulus
          signaling receptor binding
          structural molecule activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P752336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721224 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108870 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P752336 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P752336 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P752336 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P752336 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46727 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10784811 - -
          Affymetrix Rat Gene 1.0 ST Array 10784813 - -
          Affymetrix Rat Gene 1.0 ST Array 10784810 - -
          Affymetrix Rat Gene 1.0 ST Array 10784812 - -
          Affymetrix Rat Gene 1.1 ST Array 10784809 - -
          Rat Genome 230 2.0 Array 1367845_PM_at - -
          Rat Genome 230 2.0 Array 1367845_at - -
          Affymetrix HT Rat Focus Array 1367845_PM_at - -
          Affymetrix HT Rat Focus Array 1367845_at - -
          Affymetrix Rat Clariom S Assay PSR1500012756.rn.1 TC1500001884.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500012755.rn.1 TC1500001884.rn.1 RTA TC
          Rat Expression Set 230 1367845_at - -
          Rat Expression Set 230 1367845_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10784809 - -
          Rat Genome U34 Set Z12152_at - -
          Rat Genome U34 Set Z12152_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22333362 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370410 - -
          RatRef-12 Expression BeadChip ILMN_68492 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc