Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00567528_m1
          See other AK4 GE Assays ›
          Gene Symbol:
          Ak4
          Gene Name:
          adenylate kinase 4
          Gene Aliases:
          AK 4, Ak3l1, Ak3l2
          Chromosome Location:
          Chr.5: 120250306 - 120309153 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00567528_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ak4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017135.3 NP_058831.1 3 - 4 438 71
          XM_006238445.2 XP_006238507.1 4 - 5 743 71
          GenBank mRNA BC087024.1 - 3 - 4 490 71
          D87809.1 - 3 - 4 438 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001073.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ak4

          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          nucleoside diphosphate phosphorylation
          ADP biosynthetic process
          brain development
          nucleoside triphosphate biosynthetic process
          response to xenobiotic stimulus
          AMP metabolic process
          ATP metabolic process
          GTP metabolic process
          molecular_function
          adenylate kinase activity
          nucleoside diphosphate kinase activity
          ATP binding
          GTP binding
          nucleoside triphosphate adenylate kinase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11831 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P745269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880199 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48045 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77920 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28314 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58746 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28771 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167749 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P745269 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11831 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11831 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P745269 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P745269 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11831 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P745269 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11831 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19498 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71600 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94780 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45529 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10870268 - -
          Affymetrix Rat Gene 1.0 ST Array 10870267 - -
          Affymetrix Rat Gene 1.0 ST Array 10870265 - -
          Affymetrix Rat Gene 1.0 ST Array 10870266 - -
          Affymetrix Rat Gene 1.0 ST Array 10870264 - -
          Affymetrix Rat Gene 1.0 ST Array 10870262 - -
          Affymetrix Rat Gene 1.0 ST Array 10870263 - -
          Affymetrix Rat Gene 1.1 ST Array 10870261 - -
          Rat Genome 230 2.0 Array 1371824_at - -
          Rat Genome 230 2.0 Array 1398285_at - -
          Rat Genome 230 2.0 Array 1398285_PM_at - -
          Rat Genome 230 2.0 Array 1371824_PM_at - -
          Affymetrix HT Rat Focus Array 1371824_PM_at - -
          Affymetrix HT Rat Focus Array 1398285_at - -
          Affymetrix HT Rat Focus Array 1398285_PM_at - -
          Affymetrix HT Rat Focus Array 1371824_at - -
          Affymetrix Rat Clariom S Assay PSR0500006880.rn.1 TC0500001073.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500006881.rn.1 TC0500001073.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500006882.rn.1 TC0500001073.rn.1 RTA TC
          Rat Expression Set 230 1398285_PM_at - -
          Rat Expression Set 230 1371824_PM_at - -
          Rat Expression Set 230 1398285_at - -
          Rat Expression Set 230 1371824_at - -
          Affymetrix Rat Gene 1.0 ST Array 10870261 - -
          Rat Genome U34 Set rc_AA891949_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20799582 - -
          ABI Rat Genome Survey Microarray 21861648 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375179 - -
          RatRef-12 Expression BeadChip ILMN_54512 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0