Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00567974_m1
          See other TRPV2 GE Assays ›
          Gene Symbol:
          Trpv2
          Gene Name:
          transient receptor potential cation channel, subfamily V, member 2
          Gene Aliases:
          Vrl1
          Chromosome Location:
          Chr.10: 48903540 - 48925036 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00567974_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trpv2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001270797.1 NP_001257726.1 2 - 3 583 98
          NM_001270798.1 NP_001257727.1 2 - 3 522 98
          NM_017207.3 NP_058903.2 2 - 3 606 98
          GenBank mRNA AB022332.1 - 2 - 3 538 98
          AB029330.1 - 2 - 3 505 98
          AF129113.1 - 2 - 3 525 98
          BC089215.1 - 2 - 3 538 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000806.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trpv2

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          response to temperature stimulus
          response to heat
          positive regulation of axon extension
          calcium ion transmembrane transport
          positive regulation of calcium ion import
          cation channel activity
          calcium channel activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P130184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442292 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101884 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34320 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104541 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P130184 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P130184 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P130184 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P816020 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57416 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25416 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10734333 - -
          Affymetrix Rat Gene 1.0 ST Array 10734332 - -
          Affymetrix Rat Gene 1.0 ST Array 10734321 - -
          Affymetrix Rat Gene 1.0 ST Array 10734336 - -
          Affymetrix Rat Gene 1.0 ST Array 10734330 - -
          Affymetrix Rat Gene 1.0 ST Array 10734326 - -
          Affymetrix Rat Gene 1.0 ST Array 10734322 - -
          Affymetrix Rat Gene 1.0 ST Array 10734325 - -
          Affymetrix Rat Gene 1.0 ST Array 10734329 - -
          Affymetrix Rat Gene 1.0 ST Array 10734327 - -
          Affymetrix Rat Gene 1.0 ST Array 10734323 - -
          Affymetrix Rat Gene 1.0 ST Array 10734328 - -
          Affymetrix Rat Gene 1.0 ST Array 10734334 - -
          Affymetrix Rat Gene 1.0 ST Array 10734331 - -
          Affymetrix Rat Gene 1.0 ST Array 10734324 - -
          Affymetrix Rat Gene 1.0 ST Array 10734335 - -
          Affymetrix Rat Gene 1.1 ST Array 10734320 - -
          Rat Genome 230 2.0 Array 1368735_PM_a_at - -
          Rat Genome 230 2.0 Array 1368735_a_at - -
          Affymetrix HT Rat Focus Array 1368735_a_at - -
          Affymetrix HT Rat Focus Array 1368735_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR1000006792.rn.1 TC1000000806.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000006791.rn.1 TC1000000806.rn.1 RTA TC
          Rat Expression Set 230 1368735_a_at - -
          Rat Expression Set 230 1368735_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10734320 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20976753 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2039689 - -
          RatRef-12 Expression BeadChip ILMN_48835 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0