Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00568183_m1
          See other LOC100911672 GE Assays ›
          Gene Symbol:
          LOC100911672 Atn1
          Gene Name:
          atrophin-1-like
          atrophin 1
          Gene Aliases:
          -
          Drpla
          Chromosome Location:
          Chr.9: 20264423 - 20278263 on Build Rnor_6.0
          Chromosome Location:
          Chr.4: 157267394 - 157281199 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn00568183_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100911672
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003750658.3 XP_003750706.1 4 - 5 588 66
          XM_006244637.2 XP_006244699.1 4 - 5 574 66
          XM_008766913.1 XP_008765135.1 4 - 5 556 66
          Gene Symbol: Atn1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA CB702622.1 - 1 - 2 113 66
          U31777.1 - 4 - 5 601 66

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P013456 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P023536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10763 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81393 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P013456 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P023536 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P013456 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P023536 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P023536 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P013456 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P516561 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103321 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53853 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10865495 - -
          Affymetrix Rat Gene 1.0 ST Array 10865489 - -
          Affymetrix Rat Gene 1.0 ST Array 10865490 - -
          Affymetrix Rat Gene 1.0 ST Array 10865501 - -
          Affymetrix Rat Gene 1.0 ST Array 10865496 - -
          Affymetrix Rat Gene 1.0 ST Array 10865494 - -
          Affymetrix Rat Gene 1.0 ST Array 10865499 - -
          Affymetrix Rat Gene 1.0 ST Array 10865491 - -
          Affymetrix Rat Gene 1.0 ST Array 10865488 - -
          Affymetrix Rat Gene 1.0 ST Array 10865500 - -
          Affymetrix Rat Gene 1.0 ST Array 10865498 - -
          Affymetrix Rat Gene 1.0 ST Array 10865492 - -
          Affymetrix Rat Gene 1.0 ST Array 10865493 - -
          Affymetrix Rat Gene 1.0 ST Array 10865497 - -
          Affymetrix Rat Gene 1.1 ST Array 10865487 - -
          Rat Genome 230 2.0 Array 1386972_PM_at - -
          Rat Genome 230 2.0 Array 1386972_at - -
          Affymetrix HT Rat Focus Array 1386972_PM_at - -
          Affymetrix HT Rat Focus Array 1386972_at - -
          Rat Expression Set 230 1386972_PM_at - -
          Rat Expression Set 230 1386972_at - -
          Affymetrix Rat Gene 1.0 ST Array 10865487 - -
          Rat Genome U34 Set U31777_at - -
          Rat Genome U34 Set U31777_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21726099 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0