Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00569045_m1
          See other ACO1 GE Assays ›
          Gene Symbol:
          Aco1
          Gene Name:
          aconitase 1, soluble
          Gene Aliases:
          AH, Acon1, IRP1
          Chromosome Location:
          Chr.5: 56425076 - 56481218 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00569045_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aco1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017321.1 NP_059017.1 18 - 19 2484 57
          GenBank mRNA FQ213654.1 - 3 - 4 315 57
          L23874.1 - 18 - 19 2484 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500000467.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aco1

          Gene Ontology Categories:

          Function(s) Process(es)

          tricarboxylic acid cycle
          citrate metabolic process
          regulation of translation
          cellular iron ion homeostasis
          post-embryonic development
          response to iron(II) ion
          regulation of gene expression
          intestinal absorption
          RNA binding
          mRNA binding
          aconitate hydratase activity
          iron-responsive element binding
          metal ion binding
          mRNA 5'-UTR binding
          iron-sulfur cluster binding
          4 iron, 4 sulfur cluster binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P548118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527827 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125034 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24295 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99761 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62201 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154614 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108663 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136397 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30821 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101108 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105657 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99282 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P548118 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P548118 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P548118 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P548118 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700184291 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701726551 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700491203 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25761 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19410 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48620 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78883 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9577 - -
          Agilent Rat cDNA Microarray (G4105A) 700491203 - -
          Agilent Rat cDNA Microarray (G4105A) 701726551 - -
          Agilent Rat cDNA Microarray (G4105A) 700184291 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10868272 - -
          Affymetrix Rat Gene 1.0 ST Array 10868269 - -
          Affymetrix Rat Gene 1.0 ST Array 10868271 - -
          Affymetrix Rat Gene 1.0 ST Array 10868261 - -
          Affymetrix Rat Gene 1.0 ST Array 10868256 - -
          Affymetrix Rat Gene 1.0 ST Array 10868273 - -
          Affymetrix Rat Gene 1.0 ST Array 10868270 - -
          Affymetrix Rat Gene 1.0 ST Array 10868268 - -
          Affymetrix Rat Gene 1.0 ST Array 10868260 - -
          Affymetrix Rat Gene 1.0 ST Array 10868264 - -
          Affymetrix Rat Gene 1.0 ST Array 10868258 - -
          Affymetrix Rat Gene 1.0 ST Array 10868262 - -
          Affymetrix Rat Gene 1.0 ST Array 10868257 - -
          Affymetrix Rat Gene 1.0 ST Array 10868266 - -
          Affymetrix Rat Gene 1.0 ST Array 10868259 - -
          Affymetrix Rat Gene 1.0 ST Array 10868267 - -
          Affymetrix Rat Gene 1.0 ST Array 10868263 - -
          Affymetrix Rat Gene 1.0 ST Array 10868276 - -
          Affymetrix Rat Gene 1.0 ST Array 10868274 - -
          Affymetrix Rat Gene 1.0 ST Array 10868265 - -
          Affymetrix Rat Gene 1.1 ST Array 10868255 - -
          Rat Genome 230 2.0 Array 1386916_PM_at - -
          Rat Genome 230 2.0 Array 1386916_at - -
          Affymetrix HT Rat Focus Array 1386916_at - -
          Affymetrix HT Rat Focus Array 1386916_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500002791.rn.1 TC0500000467.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500002793.rn.1 TC0500000467.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500002792.rn.1 TC0500000467.rn.1 RTA TC
          Rat Expression Set 230 1386916_PM_at - -
          Rat Expression Set 230 1386916_at - -
          Affymetrix Rat Gene 1.0 ST Array 10868255 - -
          Rat Genome U34 Set rc_AI176621_at - -
          Rat Genome U34 Set rc_AI176621_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21320062 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376951 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d58dc5f8-fsfvr:80/100.66.134.75:80.
          git-commit: 38b8a393e477ae4d9d89cc5f7a29948d21ae29e2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc