Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00569753_m1
          See other CDH22 GE Assays ›
          Gene Symbol:
          Cdh22
          Gene Name:
          cadherin 22
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 161574993 - 161970872 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00569753_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdh22
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019161.2 NP_062034.2 7 - 8 1680 137
          GenBank mRNA D83348.1 - 7 - 8 1686 137

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300004770.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdh22

          Gene Ontology Categories:

          Function(s) Process(es)

          homophilic cell adhesion via plasma membrane adhesion molecules
          brain development
          calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P033979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820642 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6587 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134908 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163113 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160237 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53835 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94373 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51038 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46187 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26973 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50867 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165571 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15532 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123891 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70884 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143037 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P033979 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P033979 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P033979 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P454259 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P116532 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102385 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38961 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99295 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40053 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62319 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57993 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95426 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20849 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10851735 - -
          Affymetrix Rat Gene 1.0 ST Array 10851725 - -
          Affymetrix Rat Gene 1.0 ST Array 10851734 - -
          Affymetrix Rat Gene 1.0 ST Array 10851733 - -
          Affymetrix Rat Gene 1.0 ST Array 10851736 - -
          Affymetrix Rat Gene 1.0 ST Array 10851737 - -
          Affymetrix Rat Gene 1.0 ST Array 10851727 - -
          Affymetrix Rat Gene 1.0 ST Array 10851730 - -
          Affymetrix Rat Gene 1.0 ST Array 10851731 - -
          Affymetrix Rat Gene 1.0 ST Array 10851732 - -
          Affymetrix Rat Gene 1.0 ST Array 10851726 - -
          Affymetrix Rat Gene 1.0 ST Array 10851738 - -
          Affymetrix Rat Gene 1.1 ST Array 10851724 - -
          Rat Genome 230 2.0 Array 1388045_a_at - -
          Rat Genome 230 2.0 Array 1388045_PM_a_at - -
          Rat Genome 230 2.0 Array 1368887_PM_at - -
          Rat Genome 230 2.0 Array 1368887_at - -
          Affymetrix HT Rat Focus Array 1368887_PM_at - -
          Affymetrix HT Rat Focus Array 1388045_a_at - -
          Affymetrix HT Rat Focus Array 1368887_at - -
          Affymetrix HT Rat Focus Array 1388045_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR0300036556.rn.1 TC0300004770.rn.1 RTA TC
          Rat Expression Set 230 1388045_PM_a_at - -
          Rat Expression Set 230 1388045_a_at - -
          Rat Expression Set 230 1368887_PM_at - -
          Rat Expression Set 230 1368887_at - -
          Affymetrix Rat Gene 1.0 ST Array 10851724 - -
          Rat Genome U34 Set D83348_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21633870 - -
          Array Name Probe IDs Targets Target Type
          RatRef-12 Expression BeadChip ILMN_50959 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0