Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00571066_m1
          See other RGS8 GE Assays ›
          Gene Symbol:
          Rgs8
          Gene Name:
          regulator of G-protein signaling 8
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 71086711 - 71141820 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00571066_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rgs8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019344.2 NP_062217.1 6 - 7 767 68
          XM_006250015.2 XP_006250077.1 5 - 6 638 68
          XM_006250016.2 XP_006250078.1 6 - 7 582 68
          XM_006250017.2 XP_006250079.1 6 - 7 932 68
          XM_008769675.1 XP_008767897.1 6 - 7 933 68
          XM_008769676.1 XP_008767898.1 6 - 7 1098 68
          GenBank mRNA AB006013.1 - 5 - 6 676 68
          BC089064.1 - 6 - 7 767 68
          U32432.1 - 1 - 2 104 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000494.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rgs8

          Gene Ontology Categories:

          Function(s) Process(es)

          termination of G protein-coupled receptor signaling pathway
          positive regulation of GTPase activity
          GTPase activator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P465953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840748 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9772 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57473 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69078 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P465953 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P465953 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P465953 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P465953 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701896338 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79205 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14178 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97528 - -
          Agilent Rat cDNA Microarray (G4105A) 701896338 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10764765 - -
          Affymetrix Rat Gene 1.0 ST Array 10764767 - -
          Affymetrix Rat Gene 1.0 ST Array 10764766 - -
          Affymetrix Rat Gene 1.0 ST Array 10764764 - -
          Affymetrix Rat Gene 1.0 ST Array 10764768 - -
          Affymetrix Rat Gene 1.0 ST Array 10764762 - -
          Affymetrix Rat Gene 1.0 ST Array 10764769 - -
          Affymetrix Rat Gene 1.0 ST Array 10764763 - -
          Affymetrix Rat Gene 1.1 ST Array 10764761 - -
          Rat Genome 230 2.0 Array 1387403_PM_at - -
          Rat Genome 230 2.0 Array 1387403_at - -
          Affymetrix HT Rat Focus Array 1387403_at - -
          Affymetrix HT Rat Focus Array 1387403_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300003507.rn.1 TC1300000494.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300003505.rn.1 TC1300000494.rn.1 RTA TC
          Rat Expression Set 230 1387403_at - -
          Rat Expression Set 230 1387403_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10764761 - -
          Rat Genome U34 Set AB006013_at - -
          Rat Genome U34 Set rc_AI144709_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21824225 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371675 - -
          RatRef-12 Expression BeadChip ILMN_55584 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0